Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58305782-58306015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPOAP1
Entrez Gene ID
9256
Gene Name
TSPO associated protein 1
Gene Aliases
BZRAP1, PBR-IP, PRAX-1, PRAX1, RIM-BP1, RIMBP1
UniGene
Hs.112499
Gene Chromosome Location
Chr.17: 58301227-58329030 on Build GRCh38
Transcripts
NM_001261835.1 NM_004758.3 NM_024418.2 XM_005257807.2 XM_006722173.2 XM_006722174.2 XM_006722175.2 XM_006722176.2 XM_006722177.3 XM_006722178.3 XM_011525471.2 XM_011525472.1 XM_017025326.1 XM_017025327.1 XM_017025328.1 XM_017025329.1 XM_017025330.1 XM_017025331.1 XM_017025332.1 XM_017025333.1 XM_017025334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6211636 c.5225-6C>A p.? Chr.17: 58305871-58305871 on Build GRCh38
6417146 c.5249C>G p.P1750R Chr.17: 58305841-58305841 on Build GRCh38
6211637 c.5225-6C>A p.? Chr.17: 58305871-58305871 on Build GRCh38
5041880 c.5225-1G>A p.? Chr.17: 58305866-58305866 on Build GRCh38
5041881 c.5225-1G>A p.? Chr.17: 58305866-58305866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776184072 Chr.17: 58305836-58305836 on Build GRCh38
rs563853077 Chr.17: 58305893-58305893 on Build GRCh38
rs899967193 Chr.17: 58305987-58305987 on Build GRCh38
rs528347679 Chr.17: 58305981-58305981 on Build GRCh38
rs571253880 Chr.17: 58305990-58305990 on Build GRCh38
rs376779101 Chr.17: 58305802-58305802 on Build GRCh38
rs760855119 Chr.17: 58305883-58305883 on Build GRCh38
rs749654267 Chr.17: 58305813-58305813 on Build GRCh38
rs546925647 Chr.17: 58305986-58305986 on Build GRCh38
rs751814072 Chr.17: 58305864-58305864 on Build GRCh38
rs779201128 Chr.17: 58305913-58305913 on Build GRCh38
rs190385470 Chr.17: 58305909-58305909 on Build GRCh38
rs369426839 Chr.17: 58305811-58305811 on Build GRCh38
rs542792801 Chr.17: 58305820-58305820 on Build GRCh38
rs370997723 Chr.17: 58305805-58305805 on Build GRCh38
rs754148476 Chr.17: 58305946-58305946 on Build GRCh38
rs142624026 Chr.17: 58305844-58305844 on Build GRCh38
rs1047288069 Chr.17: 58305950-58305950 on Build GRCh38
rs139473278 Chr.17: 58305858-58305858 on Build GRCh38
rs895644504 Chr.17: 58305894-58305894 on Build GRCh38
rs368096909 Chr.17: 58305810-58305810 on Build GRCh38
rs757563585 Chr.17: 58305816-58305816 on Build GRCh38
rs752821231 Chr.17: 58305866-58305866 on Build GRCh38
rs567490250 Chr.17: 58305922-58305922 on Build GRCh38
rs996879749 Chr.17: 58305994-58305994 on Build GRCh38
rs756210553 Chr.17: 58305884-58305884 on Build GRCh38
rs148464443 Chr.17: 58305843-58305843 on Build GRCh38
rs766233835 Chr.17: 58305846-58305846 on Build GRCh38
rs768320600 Chr.17: 58305832-58305832 on Build GRCh38
rs372277686 Chr.17: 58305898-58305898 on Build GRCh38
rs747678697 Chr.17: 58305839-58305839 on Build GRCh38
rs781638492 Chr.17: 58305980-58305980 on Build GRCh38
rs1036323089 Chr.17: 58305931-58305931 on Build GRCh38
rs3744102 Chr.17: 58305826-58305826 on Build GRCh38
rs774307691 Chr.17: 58305857-58305857 on Build GRCh38
rs557358633 Chr.17: 58305819-58305819 on Build GRCh38
rs908552521 Chr.17: 58305958-58305958 on Build GRCh38
rs946699944 Chr.17: 58305828-58305828 on Build GRCh38
rs779256006 Chr.17: 58305824-58305824 on Build GRCh38

View Full Product Details