Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17499369-17499630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR2
Entrez Gene ID
27443
Gene Name
CECR2, histone acetyl-lysine reader
Gene Aliases
-
UniGene
Hs.231895
Gene Chromosome Location
Chr.22: 17359949-17558155 on Build GRCh38
Transcripts
NM_001290046.1 NM_001290047.1 XM_011546128.2 XM_011546129.2 XM_011546132.2 XM_017028785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
478704 c.392G>T p.G131V Chr.22: 17499462-17499462 on Build GRCh38
7147163 c.422G>A p.R141Q Chr.22: 17499492-17499492 on Build GRCh38
4102410 c.361C>T p.R121C Chr.22: 17499431-17499431 on Build GRCh38
7147164 c.362G>A p.R121Q Chr.22: 17499492-17499492 on Build GRCh38
4102407 c.347A>G p.D116G Chr.22: 17499417-17499417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367590870 Chr.22: 17499510-17499510 on Build GRCh38
rs759014846 Chr.22: 17499494-17499494 on Build GRCh38
rs549635868 Chr.22: 17499467-17499467 on Build GRCh38
rs536039455 Chr.22: 17499442-17499442 on Build GRCh38
rs751690552 Chr.22: 17499506-17499506 on Build GRCh38
rs749039356 Chr.22: 17499396-17499396 on Build GRCh38
rs768318389 Chr.22: 17499539-17499539 on Build GRCh38
rs773302104 Chr.22: 17499564-17499564 on Build GRCh38
rs7284169 Chr.22: 17499568-17499568 on Build GRCh38
rs765519450 Chr.22: 17499599-17499599 on Build GRCh38
rs768499585 Chr.22: 17499397-17499397 on Build GRCh38
rs772888002 Chr.22: 17499443-17499443 on Build GRCh38
rs781322518 Chr.22: 17499520-17499520 on Build GRCh38
rs760380592 Chr.22: 17499456-17499456 on Build GRCh38
rs1031496252 Chr.22: 17499400-17499400 on Build GRCh38
rs753135447 Chr.22: 17499469-17499469 on Build GRCh38
rs776365846 Chr.22: 17499591-17499591 on Build GRCh38
rs772094030 Chr.22: 17499557-17499557 on Build GRCh38
rs746099602 Chr.22: 17499528-17499528 on Build GRCh38
rs779804126 Chr.22: 17499390-17499390 on Build GRCh38
rs774093736 Chr.22: 17499413-17499413 on Build GRCh38
rs761864121 Chr.22: 17499421-17499421 on Build GRCh38
rs771185505 Chr.22: 17499388-17499388 on Build GRCh38
rs753577889 Chr.22: 17499463-17499463 on Build GRCh38
rs989988254 Chr.22: 17499580-17499580 on Build GRCh38
rs777687239 Chr.22: 17499503-17499503 on Build GRCh38
rs759291343 Chr.22: 17499597-17499597 on Build GRCh38
rs748003540 Chr.22: 17499554-17499554 on Build GRCh38
rs770571521 Chr.22: 17499569-17499569 on Build GRCh38
rs370206086 Chr.22: 17499594-17499594 on Build GRCh38
rs766083330 Chr.22: 17499457-17499457 on Build GRCh38
rs535340689 Chr.22: 17499552-17499552 on Build GRCh38

View Full Product Details