Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176117314-176117559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD10
Entrez Gene ID
3236
Gene Name
homeobox D10
Gene Aliases
HOX4, HOX4D, HOX4E, Hox-4.4
UniGene
Hs.123070
Gene Chromosome Location
Chr.2: 176116764-176119942 on Build GRCh38
Transcripts
NM_002148.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4777286 c.679G>C p.E227Q Chr.2: 176117512-176117512 on Build GRCh38
6510039 c.627G>A p.E209E Chr.2: 176117460-176117460 on Build GRCh38
4691027 c.613G>A p.V205M Chr.2: 176117446-176117446 on Build GRCh38
3570192 c.625G>A p.E209K Chr.2: 176117458-176117458 on Build GRCh38
3570191 c.600G>T p.K200N Chr.2: 176117433-176117433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2593777 Chr.2: 176117397-176117397 on Build GRCh38
rs748360990 Chr.2: 176117343-176117343 on Build GRCh38
rs147270324 Chr.2: 176117420-176117420 on Build GRCh38
rs201752392 Chr.2: 176117494-176117494 on Build GRCh38
rs763788485 Chr.2: 176117380-176117380 on Build GRCh38
rs377417077 Chr.2: 176117503-176117503 on Build GRCh38
rs1046890556 Chr.2: 176117378-176117378 on Build GRCh38
rs759998982 Chr.2: 176117434-176117434 on Build GRCh38
rs1052405205 Chr.2: 176117493-176117493 on Build GRCh38
rs148563899 Chr.2: 176117452-176117452 on Build GRCh38
rs575171191 Chr.2: 176117456-176117456 on Build GRCh38
rs886055158 Chr.2: 176117376-176117376 on Build GRCh38
rs758327405 Chr.2: 176117403-176117403 on Build GRCh38
rs779016325 Chr.2: 176117491-176117491 on Build GRCh38
rs778517437 Chr.2: 176117386-176117386 on Build GRCh38
rs112907565 Chr.2: 176117531-176117531 on Build GRCh38
rs754834998 Chr.2: 176117541-176117541 on Build GRCh38
rs770037719 Chr.2: 176117344-176117344 on Build GRCh38
rs749450422 Chr.2: 176117419-176117419 on Build GRCh38
rs560628362 Chr.2: 176117490-176117490 on Build GRCh38
rs143870330 Chr.2: 176117394-176117394 on Build GRCh38
rs749929272 Chr.2: 176117453-176117453 on Build GRCh38
rs765744983 Chr.2: 176117526-176117526 on Build GRCh38
rs563184541 Chr.2: 176117444-176117444 on Build GRCh38
rs143111542 Chr.2: 176117439-176117439 on Build GRCh38
rs762526013 Chr.2: 176117521-176117521 on Build GRCh38
rs761097733 Chr.2: 176117441-176117441 on Build GRCh38
rs868138731 Chr.2: 176117389-176117389 on Build GRCh38
rs764879434 Chr.2: 176117446-176117446 on Build GRCh38
rs753072399 Chr.2: 176117371-176117371 on Build GRCh38
rs980924218 Chr.2: 176117433-176117433 on Build GRCh38
rs769981285 Chr.2: 176117414-176117414 on Build GRCh38
rs773432025 Chr.2: 176117415-176117415 on Build GRCh38
rs753465255 Chr.2: 176117381-176117381 on Build GRCh38
rs1052313791 Chr.2: 176117396-176117396 on Build GRCh38
rs868004054 Chr.2: 176117512-176117512 on Build GRCh38
rs777103841 Chr.2: 176117509-176117509 on Build GRCh38
rs916675788 Chr.2: 176117349-176117349 on Build GRCh38
rs759983428 Chr.2: 176117362-176117362 on Build GRCh38
rs773340507 Chr.2: 176117351-176117351 on Build GRCh38
rs754811472 Chr.2: 176117466-176117466 on Build GRCh38
rs746808808 Chr.2: 176117407-176117407 on Build GRCh38
rs751382680 Chr.2: 176117464-176117464 on Build GRCh38
rs775316778 Chr.2: 176117342-176117342 on Build GRCh38
rs776070838 Chr.2: 176117501-176117501 on Build GRCh38
rs2593778 Chr.2: 176117398-176117398 on Build GRCh38
rs372134472 Chr.2: 176117359-176117359 on Build GRCh38
rs758323081 Chr.2: 176117455-176117455 on Build GRCh38
rs749965639 Chr.2: 176117519-176117519 on Build GRCh38
rs763020445 Chr.2: 176117352-176117352 on Build GRCh38
rs542394792 Chr.2: 176117470-176117470 on Build GRCh38
rs756832722 Chr.2: 176117385-176117385 on Build GRCh38
rs756929170 Chr.2: 176117445-176117445 on Build GRCh38
rs767800637 Chr.2: 176117367-176117367 on Build GRCh38
rs766383927 Chr.2: 176117356-176117356 on Build GRCh38
rs755769889 Chr.2: 176117374-176117374 on Build GRCh38
rs774847930 Chr.2: 176117361-176117361 on Build GRCh38
rs775774991 Chr.2: 176117440-176117440 on Build GRCh38
rs745832332 Chr.2: 176117395-176117395 on Build GRCh38
rs749546722 Chr.2: 176117477-176117477 on Build GRCh38
rs769173252 Chr.2: 176117504-176117504 on Build GRCh38
rs939576278 Chr.2: 176117479-176117479 on Build GRCh38
rs764487159 Chr.2: 176117443-176117443 on Build GRCh38
rs151123199 Chr.2: 176117511-176117511 on Build GRCh38

View Full Product Details