Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87196603-87196872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM243
Entrez Gene ID
79161
Gene Name
transmembrane protein 243
Gene Aliases
C7orf23, MM-TRAG, MMTRAG
UniGene
Hs.619593
Gene Chromosome Location
Chr.7: 87195593-87220603 on Build GRCh38
Transcripts
XM_005250589.3 XM_017012617.1 XM_017012618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7246979 c.343G>A p.D115N Chr.7: 87196650-87196650 on Build GRCh38
453496 c.348G>A p.V116V Chr.7: 87196645-87196645 on Build GRCh38
7294570 c.258C>A p.D86E Chr.7: 87196735-87196735 on Build GRCh38
1092553 c.267G>A p.P89P Chr.7: 87196726-87196726 on Build GRCh38
7135863 c.256G>C p.D86H Chr.7: 87196737-87196737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369528589 Chr.7: 87196800-87196800 on Build GRCh38
rs142491708 Chr.7: 87196674-87196674 on Build GRCh38
rs375982092 Chr.7: 87196745-87196745 on Build GRCh38
rs189574578 Chr.7: 87196843-87196843 on Build GRCh38
rs778815607 Chr.7: 87196768-87196768 on Build GRCh38
rs1038582170 Chr.7: 87196699-87196699 on Build GRCh38
rs41278755 Chr.7: 87196759-87196759 on Build GRCh38
rs775096609 Chr.7: 87196681-87196681 on Build GRCh38
rs748967954 Chr.7: 87196672-87196672 on Build GRCh38
rs184059498 Chr.7: 87196737-87196737 on Build GRCh38
rs764555050 Chr.7: 87196634-87196634 on Build GRCh38
rs745543726 Chr.7: 87196779-87196779 on Build GRCh38
rs748125266 Chr.7: 87196661-87196661 on Build GRCh38
rs148829746 Chr.7: 87196664-87196664 on Build GRCh38
rs771744709 Chr.7: 87196678-87196678 on Build GRCh38
rs765313793 Chr.7: 87196642-87196642 on Build GRCh38
rs540968065 Chr.7: 87196847-87196847 on Build GRCh38
rs994415618 Chr.7: 87196754-87196754 on Build GRCh38
rs768990718 Chr.7: 87196698-87196698 on Build GRCh38
rs779728640 Chr.7: 87196802-87196802 on Build GRCh38
rs780046076 Chr.7: 87196763-87196765 on Build GRCh38
rs757048271 Chr.7: 87196760-87196760 on Build GRCh38
rs780921246 Chr.7: 87196629-87196630 on Build GRCh38
rs907562581 Chr.7: 87196821-87196821 on Build GRCh38
rs780085545 Chr.7: 87196653-87196653 on Build GRCh38
rs766468075 Chr.7: 87196727-87196727 on Build GRCh38
rs751580444 Chr.7: 87196733-87196733 on Build GRCh38
rs750694074 Chr.7: 87196716-87196716 on Build GRCh38
rs950098917 Chr.7: 87196649-87196649 on Build GRCh38
rs373365656 Chr.7: 87196633-87196633 on Build GRCh38
rs745666137 Chr.7: 87196752-87196754 on Build GRCh38
rs745490758 Chr.7: 87196677-87196677 on Build GRCh38
rs940351491 Chr.7: 87196835-87196835 on Build GRCh38
rs755808865 Chr.7: 87196761-87196762 on Build GRCh38
rs906583689 Chr.7: 87196635-87196635 on Build GRCh38
rs78040532 Chr.7: 87196697-87196697 on Build GRCh38
rs775600302 Chr.7: 87196774-87196775 on Build GRCh38
rs777636881 Chr.7: 87196668-87196668 on Build GRCh38
rs763131561 Chr.7: 87196726-87196726 on Build GRCh38
rs768302736 Chr.7: 87196797-87196798 on Build GRCh38
rs770753289 Chr.7: 87196673-87196673 on Build GRCh38
rs765623504 Chr.7: 87196705-87196705 on Build GRCh38
rs762154840 Chr.7: 87196704-87196704 on Build GRCh38
rs368911823 Chr.7: 87196703-87196703 on Build GRCh38
rs375970235 Chr.7: 87196646-87196646 on Build GRCh38
rs754107238 Chr.7: 87196637-87196637 on Build GRCh38
rs750641077 Chr.7: 87196644-87196644 on Build GRCh38
rs757517600 Chr.7: 87196640-87196640 on Build GRCh38

View Full Product Details