Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 88143776-88144015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNR1
Entrez Gene ID
1268
Gene Name
cannabinoid receptor 1
Gene Aliases
CANN6, CB-R, CB1, CB1A, CB1K5, CB1R, CNR
UniGene
Hs.75110
Gene Chromosome Location
Chr.6: 88139864-88167429 on Build GRCh38
Transcripts
NM_001160226.1 NM_001160258.1 NM_001160259.1 NM_016083.4 NM_033181.3 XM_006715330.3 XM_011535424.1 XM_011535425.1 XM_011535426.1 XM_011535428.1 XM_017010238.1 XM_017010239.1 XM_017010240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6047560 c.1271A>T p.N424I Chr.6: 88144004-88144004 on Build GRCh38
6631603 c.1336G>A p.A446T Chr.6: 88143939-88143939 on Build GRCh38
5436123 c.1400C>T p.T467M Chr.6: 88143875-88143875 on Build GRCh38
1082391 c.1358C>T p.T453M Chr.6: 88143917-88143917 on Build GRCh38
4153647 c.1369G>T p.A457S Chr.6: 88143906-88143906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773947953 Chr.6: 88143890-88143890 on Build GRCh38
rs946462817 Chr.6: 88143932-88143932 on Build GRCh38
rs563012512 Chr.6: 88143825-88143825 on Build GRCh38
rs772744225 Chr.6: 88143811-88143811 on Build GRCh38
rs770665729 Chr.6: 88143986-88143986 on Build GRCh38
rs778223069 Chr.6: 88143811-88143812 on Build GRCh38
rs767998702 Chr.6: 88143913-88143913 on Build GRCh38
rs370487985 Chr.6: 88143969-88143969 on Build GRCh38
rs753068397 Chr.6: 88143833-88143833 on Build GRCh38
rs200738920 Chr.6: 88143830-88143830 on Build GRCh38
rs1049353 Chr.6: 88143916-88143916 on Build GRCh38
rs768591630 Chr.6: 88143864-88143864 on Build GRCh38
rs546304911 Chr.6: 88143809-88143809 on Build GRCh38
rs75641625 Chr.6: 88143970-88143970 on Build GRCh38
rs769446052 Chr.6: 88143823-88143825 on Build GRCh38
rs769601091 Chr.6: 88143972-88143972 on Build GRCh38
rs766054760 Chr.6: 88143823-88143823 on Build GRCh38
rs772492894 Chr.6: 88143939-88143939 on Build GRCh38
rs745517223 Chr.6: 88143812-88143813 on Build GRCh38
rs749116792 Chr.6: 88143983-88143983 on Build GRCh38
rs762494727 Chr.6: 88143812-88143812 on Build GRCh38
rs777759836 Chr.6: 88143973-88143973 on Build GRCh38
rs760079776 Chr.6: 88143917-88143917 on Build GRCh38
rs141056981 Chr.6: 88143909-88143909 on Build GRCh38
rs750464422 Chr.6: 88143945-88143945 on Build GRCh38
rs779008264 Chr.6: 88143859-88143859 on Build GRCh38
rs747911868 Chr.6: 88143871-88143871 on Build GRCh38
rs376916854 Chr.6: 88143940-88143940 on Build GRCh38
rs138027855 Chr.6: 88143875-88143875 on Build GRCh38
rs762733641 Chr.6: 88143883-88143883 on Build GRCh38
rs776465774 Chr.6: 88143868-88143868 on Build GRCh38
rs755592434 Chr.6: 88143826-88143826 on Build GRCh38
rs149238893 Chr.6: 88143988-88143988 on Build GRCh38
rs1053817320 Chr.6: 88143980-88143980 on Build GRCh38
rs756443377 Chr.6: 88143836-88143836 on Build GRCh38
rs199942265 Chr.6: 88143874-88143874 on Build GRCh38
rs951061482 Chr.6: 88143927-88143927 on Build GRCh38
rs1016906543 Chr.6: 88143837-88143837 on Build GRCh38
rs746929786 Chr.6: 88143862-88143862 on Build GRCh38
rs758887693 Chr.6: 88143812-88143812 on Build GRCh38
rs765964116 Chr.6: 88143887-88143887 on Build GRCh38
rs77698129 Chr.6: 88143855-88143855 on Build GRCh38
rs764901348 Chr.6: 88143806-88143806 on Build GRCh38
rs777997554 Chr.6: 88143844-88143844 on Build GRCh38
rs749565225 Chr.6: 88143851-88143851 on Build GRCh38
rs756483875 Chr.6: 88143920-88143920 on Build GRCh38
rs764437215 Chr.6: 88143926-88143926 on Build GRCh38
rs759146606 Chr.6: 88143901-88143901 on Build GRCh38
rs758413605 Chr.6: 88143950-88143950 on Build GRCh38
rs982410039 Chr.6: 88143958-88143958 on Build GRCh38
rs375652364 Chr.6: 88143900-88143900 on Build GRCh38

View Full Product Details