Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75435434-75435695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD5
Entrez Gene ID
81544
Gene Name
glycerophosphodiester phosphodiesterase domain containing 5
Gene Aliases
GDE2, PP1665
UniGene
Hs.503297
Gene Chromosome Location
Chr.11: 75434640-75525916 on Build GRCh38
Transcripts
NM_030792.6 XM_006718697.3 XM_011545276.2 XM_011545277.2 XM_011545278.2 XM_011545279.2 XM_011545280.1 XM_011545281.2 XM_011545285.2 XM_011545286.1 XM_011545287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6670402 c.1707C>T p.S569S Chr.11: 75435618-75435618 on Build GRCh38
931988 c.1763G>A p.G588D Chr.11: 75435562-75435562 on Build GRCh38
4036735 c.1768C>T p.R590* Chr.11: 75435557-75435557 on Build GRCh38
4686753 c.1813C>T p.R605C Chr.11: 75435512-75435512 on Build GRCh38
211801 c.1787C>A p.T596N Chr.11: 75435538-75435538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983737566 Chr.11: 75435648-75435648 on Build GRCh38
rs376398826 Chr.11: 75435641-75435641 on Build GRCh38
rs749608394 Chr.11: 75435629-75435629 on Build GRCh38
rs754023687 Chr.11: 75435663-75435663 on Build GRCh38
rs778938510 Chr.11: 75435591-75435591 on Build GRCh38
rs760729755 Chr.11: 75435660-75435660 on Build GRCh38
rs750661900 Chr.11: 75435497-75435497 on Build GRCh38
rs148544864 Chr.11: 75435618-75435618 on Build GRCh38
rs752634222 Chr.11: 75435671-75435671 on Build GRCh38
rs569699868 Chr.11: 75435575-75435575 on Build GRCh38
rs750559361 Chr.11: 75435557-75435557 on Build GRCh38
rs906047811 Chr.11: 75435480-75435480 on Build GRCh38
rs1018622681 Chr.11: 75435567-75435567 on Build GRCh38
rs183534412 Chr.11: 75435458-75435458 on Build GRCh38
rs775901461 Chr.11: 75435537-75435537 on Build GRCh38
rs765157828 Chr.11: 75435670-75435670 on Build GRCh38
rs752073631 Chr.11: 75435547-75435547 on Build GRCh38
rs187459718 Chr.11: 75435624-75435624 on Build GRCh38
rs770587553 Chr.11: 75435617-75435617 on Build GRCh38
rs537176106 Chr.11: 75435630-75435630 on Build GRCh38
rs779850795 Chr.11: 75435603-75435604 on Build GRCh38
rs751633617 Chr.11: 75435485-75435485 on Build GRCh38
rs764635820 Chr.11: 75435481-75435481 on Build GRCh38
rs991810972 Chr.11: 75435483-75435483 on Build GRCh38
rs747652235 Chr.11: 75435513-75435513 on Build GRCh38
rs758166903 Chr.11: 75435470-75435471 on Build GRCh38
rs748104045 Chr.11: 75435593-75435593 on Build GRCh38
rs964545542 Chr.11: 75435568-75435568 on Build GRCh38
rs759730108 Chr.11: 75435669-75435669 on Build GRCh38
rs746550127 Chr.11: 75435609-75435609 on Build GRCh38
rs528344017 Chr.11: 75435490-75435490 on Build GRCh38
rs781609091 Chr.11: 75435551-75435551 on Build GRCh38
rs754882301 Chr.11: 75435588-75435588 on Build GRCh38
rs551593132 Chr.11: 75435511-75435511 on Build GRCh38
rs768603832 Chr.11: 75435512-75435512 on Build GRCh38
rs372960103 Chr.11: 75435542-75435542 on Build GRCh38
rs756334817 Chr.11: 75435561-75435561 on Build GRCh38
rs1027564886 Chr.11: 75435598-75435598 on Build GRCh38
rs761812654 Chr.11: 75435549-75435549 on Build GRCh38
rs770006144 Chr.11: 75435526-75435526 on Build GRCh38
rs777367424 Chr.11: 75435608-75435608 on Build GRCh38
rs746911586 Chr.11: 75435676-75435677 on Build GRCh38
rs144731330 Chr.11: 75435576-75435576 on Build GRCh38
rs757385377 Chr.11: 75435489-75435489 on Build GRCh38
rs779919308 Chr.11: 75435509-75435509 on Build GRCh38
rs764570484 Chr.11: 75435544-75435544 on Build GRCh38
rs917693526 Chr.11: 75435515-75435515 on Build GRCh38
rs745910762 Chr.11: 75435520-75435520 on Build GRCh38
rs781332651 Chr.11: 75435496-75435496 on Build GRCh38
rs766614363 Chr.11: 75435662-75435662 on Build GRCh38
rs757942561 Chr.11: 75435597-75435597 on Build GRCh38
rs767614808 Chr.11: 75435556-75435556 on Build GRCh38
rs373412770 Chr.11: 75435649-75435649 on Build GRCh38
rs201881925 Chr.11: 75435521-75435521 on Build GRCh38
rs369743472 Chr.11: 75435647-75435647 on Build GRCh38

View Full Product Details