Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157342726-157342957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYFIP2
Entrez Gene ID
26999
Gene Name
cytoplasmic FMR1 interacting protein 2
Gene Aliases
PIR121
UniGene
Hs.519702
Gene Chromosome Location
Chr.5: 157266080-157395598 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1 NM_001001343.3

Gene Symbol
FNDC9
Entrez Gene ID
408263
Gene Name
fibronectin type III domain containing 9
Gene Aliases
C5orf40
UniGene
Hs.437066
Gene Chromosome Location
Chr.5: 157341599-157345721 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1 NM_001001343.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6170465 c.623T>C p.L208S Chr.5: 157342914-157342914 on Build GRCh38
4827734 c.614C>A p.A205E Chr.5: 157342923-157342923 on Build GRCh38
6337727 c.646C>T p.P216S Chr.5: 157342891-157342891 on Build GRCh38
231913 c.626A>C p.Q209P Chr.5: 157342911-157342911 on Build GRCh38
3853148 c.633G>T p.G211G Chr.5: 157342904-157342904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778948372 Chr.5: 157342901-157342902 on Build GRCh38
rs926724317 Chr.5: 157342911-157342911 on Build GRCh38
rs768502943 Chr.5: 157342845-157342845 on Build GRCh38
rs911628481 Chr.5: 157342806-157342806 on Build GRCh38
rs890806943 Chr.5: 157342810-157342810 on Build GRCh38
rs1007808594 Chr.5: 157342825-157342825 on Build GRCh38
rs778113223 Chr.5: 157342811-157342811 on Build GRCh38
rs200165888 Chr.5: 157342907-157342907 on Build GRCh38
rs369433912 Chr.5: 157342897-157342897 on Build GRCh38
rs778027094 Chr.5: 157342879-157342879 on Build GRCh38
rs778842992 Chr.5: 157342909-157342909 on Build GRCh38
rs977773007 Chr.5: 157342768-157342768 on Build GRCh38
rs200025973 Chr.5: 157342878-157342878 on Build GRCh38
rs779332505 Chr.5: 157342819-157342819 on Build GRCh38
rs752896523 Chr.5: 157342875-157342875 on Build GRCh38
rs750367980 Chr.5: 157342864-157342864 on Build GRCh38
rs772215216 Chr.5: 157342928-157342928 on Build GRCh38
rs899581433 Chr.5: 157342854-157342854 on Build GRCh38
rs767518510 Chr.5: 157342870-157342870 on Build GRCh38
rs966525806 Chr.5: 157342759-157342759 on Build GRCh38
rs548857088 Chr.5: 157342842-157342842 on Build GRCh38
rs143433635 Chr.5: 157342908-157342908 on Build GRCh38
rs747309879 Chr.5: 157342935-157342936 on Build GRCh38
rs771510769 Chr.5: 157342838-157342838 on Build GRCh38
rs185099179 Chr.5: 157342754-157342754 on Build GRCh38
rs761670872 Chr.5: 157342901-157342901 on Build GRCh38
rs746495560 Chr.5: 157342923-157342923 on Build GRCh38
rs771550330 Chr.5: 157342891-157342891 on Build GRCh38
rs190293152 Chr.5: 157342786-157342786 on Build GRCh38
rs200489390 Chr.5: 157342902-157342902 on Build GRCh38
rs779437169 Chr.5: 157342892-157342892 on Build GRCh38
rs376408771 Chr.5: 157342846-157342846 on Build GRCh38
rs201701167 Chr.5: 157342906-157342906 on Build GRCh38
rs768671457 Chr.5: 157342842-157342842 on Build GRCh38
rs576494565 Chr.5: 157342912-157342912 on Build GRCh38
rs750878522 Chr.5: 157342910-157342910 on Build GRCh38
rs559170564 Chr.5: 157342808-157342808 on Build GRCh38
rs780670357 Chr.5: 157342922-157342922 on Build GRCh38
rs771414260 Chr.5: 157342817-157342817 on Build GRCh38
rs746587472 Chr.5: 157342895-157342895 on Build GRCh38
rs73815845 Chr.5: 157342833-157342833 on Build GRCh38
rs1022057436 Chr.5: 157342746-157342746 on Build GRCh38
rs759622473 Chr.5: 157342868-157342868 on Build GRCh38
rs754579881 Chr.5: 157342934-157342934 on Build GRCh38
rs757500549 Chr.5: 157342888-157342888 on Build GRCh38
rs749718405 Chr.5: 157342816-157342816 on Build GRCh38
rs765245315 Chr.5: 157342849-157342849 on Build GRCh38
rs776472166 Chr.5: 157342899-157342899 on Build GRCh38
rs551729243 Chr.5: 157342801-157342801 on Build GRCh38
rs748255153 Chr.5: 157342902-157342902 on Build GRCh38
rs995325867 Chr.5: 157342863-157342863 on Build GRCh38
rs553987053 Chr.5: 157342889-157342889 on Build GRCh38
rs754179786 Chr.5: 157342884-157342884 on Build GRCh38
rs773424810 Chr.5: 157342931-157342932 on Build GRCh38

View Full Product Details