Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11378576-11378835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPOR
Entrez Gene ID
2057
Gene Name
erythropoietin receptor
Gene Aliases
EPO-R
UniGene
Hs.631624
Gene Chromosome Location
Chr.19: 11377205-11384342 on Build GRCh38
Transcripts
NM_000121.3 NR_033663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5887482 c.874G>C p.E292Q Chr.19: 11378732-11378732 on Build GRCh38
990887 c.907A>C p.N303H Chr.19: 11378699-11378699 on Build GRCh38
6395256 c.886C>A p.L296I Chr.19: 11378720-11378720 on Build GRCh38
417759 c.880G>C p.E294Q Chr.19: 11378726-11378726 on Build GRCh38
5741442 c.902A>G p.K301R Chr.19: 11378704-11378704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563298152 Chr.19: 11378748-11378748 on Build GRCh38
rs756329208 Chr.19: 11378752-11378752 on Build GRCh38
rs774415917 Chr.19: 11378676-11378676 on Build GRCh38
rs771993916 Chr.19: 11378794-11378794 on Build GRCh38
rs759181310 Chr.19: 11378679-11378679 on Build GRCh38
rs777458672 Chr.19: 11378664-11378664 on Build GRCh38
rs139756642 Chr.19: 11378746-11378746 on Build GRCh38
rs749153526 Chr.19: 11378774-11378774 on Build GRCh38
rs749060498 Chr.19: 11378668-11378668 on Build GRCh38
rs771710255 Chr.19: 11378688-11378688 on Build GRCh38
rs775003412 Chr.19: 11378692-11378692 on Build GRCh38
rs1002715714 Chr.19: 11378622-11378622 on Build GRCh38
rs745809568 Chr.19: 11378793-11378793 on Build GRCh38
rs780511755 Chr.19: 11378625-11378625 on Build GRCh38
rs770845339 Chr.19: 11378775-11378775 on Build GRCh38
rs201953069 Chr.19: 11378754-11378754 on Build GRCh38
rs905718217 Chr.19: 11378600-11378600 on Build GRCh38
rs886054192 Chr.19: 11378742-11378742 on Build GRCh38
rs766932379 Chr.19: 11378733-11378733 on Build GRCh38
rs750797567 Chr.19: 11378722-11378722 on Build GRCh38
rs750747858 Chr.19: 11378605-11378605 on Build GRCh38
rs771039560 Chr.19: 11378674-11378674 on Build GRCh38
rs373802520 Chr.19: 11378796-11378796 on Build GRCh38
rs763766126 Chr.19: 11378712-11378712 on Build GRCh38
rs200991498 Chr.19: 11378608-11378608 on Build GRCh38
rs770000988 Chr.19: 11378662-11378662 on Build GRCh38
rs369085901 Chr.19: 11378745-11378745 on Build GRCh38
rs200057444 Chr.19: 11378732-11378732 on Build GRCh38
rs930684052 Chr.19: 11378777-11378777 on Build GRCh38
rs778780691 Chr.19: 11378791-11378791 on Build GRCh38
rs774151596 Chr.19: 11378802-11378803 on Build GRCh38
rs781223562 Chr.19: 11378762-11378764 on Build GRCh38
rs371902320 Chr.19: 11378631-11378631 on Build GRCh38
rs747143790 Chr.19: 11378633-11378633 on Build GRCh38
rs752058983 Chr.19: 11378736-11378736 on Build GRCh38
rs748324034 Chr.19: 11378637-11378637 on Build GRCh38
rs374871864 Chr.19: 11378635-11378635 on Build GRCh38
rs60825798 Chr.19: 11378693-11378693 on Build GRCh38

View Full Product Details