Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 275933-276158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf96
Entrez Gene ID
140680
Gene Name
chromosome 20 open reading frame 96
Gene Aliases
dJ1103G7.2
UniGene
Hs.348112
Gene Chromosome Location
Chr.20: 270863-290778 on Build GRCh38
Transcripts
NM_080571.1 NM_153269.2 XM_006723543.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5042043 c.847G>A p.A283T Chr.20: 276047-276047 on Build GRCh38
5458818 c.1023G>A p.R341R Chr.20: 275976-275976 on Build GRCh38
7234543 c.913A>T p.I305F Chr.20: 276086-276086 on Build GRCh38
4810649 c.894C>G p.V298V Chr.20: 276000-276000 on Build GRCh38
4019713 c.992G>A p.R331Q Chr.20: 276007-276007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773509328 Chr.20: 276099-276099 on Build GRCh38
rs397740163 Chr.20: 276120-276121 on Build GRCh38
rs764556586 Chr.20: 275977-275977 on Build GRCh38
rs61354763 Chr.20: 276122-276122 on Build GRCh38
rs759121364 Chr.20: 275962-275962 on Build GRCh38
rs753988669 Chr.20: 276109-276109 on Build GRCh38
rs386811461 Chr.20: 276121-276122 on Build GRCh38
rs376609517 Chr.20: 276094-276094 on Build GRCh38
rs200889477 Chr.20: 276110-276110 on Build GRCh38
rs758268652 Chr.20: 276021-276021 on Build GRCh38
rs941900711 Chr.20: 276058-276058 on Build GRCh38
rs752035358 Chr.20: 275990-275990 on Build GRCh38
rs756968503 Chr.20: 276044-276044 on Build GRCh38
rs745458252 Chr.20: 276046-276046 on Build GRCh38
rs769346598 Chr.20: 276082-276082 on Build GRCh38
rs756500081 Chr.20: 275965-275965 on Build GRCh38
rs570488394 Chr.20: 276060-276060 on Build GRCh38
rs762280683 Chr.20: 276087-276087 on Build GRCh38
rs3842445 Chr.20: 276123-276124 on Build GRCh38
rs200251815 Chr.20: 276127-276127 on Build GRCh38
rs748802884 Chr.20: 276066-276066 on Build GRCh38
rs754072214 Chr.20: 275999-275999 on Build GRCh38
rs780790822 Chr.20: 276136-276136 on Build GRCh38
rs746867839 Chr.20: 276032-276032 on Build GRCh38
rs745877230 Chr.20: 276070-276070 on Build GRCh38
rs992202096 Chr.20: 275955-275955 on Build GRCh38
rs202179540 Chr.20: 276008-276008 on Build GRCh38
rs777686797 Chr.20: 276024-276024 on Build GRCh38
rs3827148 Chr.20: 276121-276121 on Build GRCh38
rs56774790 Chr.20: 276119-276120 on Build GRCh38
rs919614812 Chr.20: 276003-276003 on Build GRCh38
rs762073039 Chr.20: 275995-275995 on Build GRCh38
rs769440813 Chr.20: 276054-276054 on Build GRCh38
rs147591491 Chr.20: 276008-276008 on Build GRCh38
rs146492837 Chr.20: 275976-275976 on Build GRCh38
rs140973635 Chr.20: 276045-276045 on Build GRCh38
rs964337705 Chr.20: 275986-275986 on Build GRCh38
rs371876916 Chr.20: 276093-276093 on Build GRCh38
rs766455676 Chr.20: 276104-276104 on Build GRCh38
rs751409429 Chr.20: 276115-276115 on Build GRCh38
rs3827147 Chr.20: 276086-276086 on Build GRCh38
rs575839927 Chr.20: 276114-276114 on Build GRCh38
rs113419123 Chr.20: 275974-275974 on Build GRCh38
rs201883976 Chr.20: 276017-276017 on Build GRCh38
rs367958111 Chr.20: 276016-276016 on Build GRCh38
rs955765263 Chr.20: 275970-275970 on Build GRCh38
rs776122731 Chr.20: 275954-275954 on Build GRCh38
rs201373081 Chr.20: 276122-276123 on Build GRCh38
rs780128041 Chr.20: 276108-276108 on Build GRCh38
rs987070571 Chr.20: 275971-275971 on Build GRCh38
rs756250702 Chr.20: 276010-276010 on Build GRCh38

View Full Product Details