Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100089153-100089380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS6
Entrez Gene ID
10980
Gene Name
COP9 signalosome subunit 6
Gene Aliases
CSN6, MOV34-34KD
UniGene
Hs.15591
Gene Chromosome Location
Chr.7: 100088960-100092200 on Build GRCh38
Transcripts
NM_006833.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5766956 c.77-8A>G p.? Chr.7: 100089282-100089282 on Build GRCh38
3267622 c.99C>T p.C33C Chr.7: 100089312-100089312 on Build GRCh38
1132897 c.141C>G p.V47V Chr.7: 100089354-100089354 on Build GRCh38
6477908 c.77-2A>C p.? Chr.7: 100089288-100089288 on Build GRCh38
3784105 c.80T>G p.V27G Chr.7: 100089293-100089293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759665460 Chr.7: 100089272-100089272 on Build GRCh38
rs371808452 Chr.7: 100089253-100089253 on Build GRCh38
rs112097083 Chr.7: 100089287-100089287 on Build GRCh38
rs778635465 Chr.7: 100089315-100089315 on Build GRCh38
rs767862494 Chr.7: 100089277-100089277 on Build GRCh38
rs766467408 Chr.7: 100089271-100089271 on Build GRCh38
rs981614771 Chr.7: 100089213-100089213 on Build GRCh38
rs764511194 Chr.7: 100089295-100089295 on Build GRCh38
rs560041581 Chr.7: 100089200-100089200 on Build GRCh38
rs745334784 Chr.7: 100089333-100089333 on Build GRCh38
rs753478190 Chr.7: 100089301-100089301 on Build GRCh38
rs915736840 Chr.7: 100089183-100089183 on Build GRCh38
rs4729575 Chr.7: 100089250-100089250 on Build GRCh38
rs375171440 Chr.7: 100089259-100089259 on Build GRCh38
rs780113727 Chr.7: 100089351-100089351 on Build GRCh38
rs189921210 Chr.7: 100089209-100089209 on Build GRCh38
rs756440179 Chr.7: 100089294-100089294 on Build GRCh38
rs942187605 Chr.7: 100089251-100089251 on Build GRCh38
rs753252878 Chr.7: 100089288-100089288 on Build GRCh38
rs1032204134 Chr.7: 100089347-100089347 on Build GRCh38
rs928255610 Chr.7: 100089220-100089220 on Build GRCh38
rs949271193 Chr.7: 100089237-100089237 on Build GRCh38
rs756804877 Chr.7: 100089312-100089312 on Build GRCh38
rs895821090 Chr.7: 100089233-100089233 on Build GRCh38
rs757945167 Chr.7: 100089342-100089342 on Build GRCh38

View Full Product Details