Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238127589-238127852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPNL
Entrez Gene ID
339768
Gene Name
espin-like
Gene Aliases
-
UniGene
Hs.127724
Gene Chromosome Location
Chr.2: 238100310-238133287 on Build GRCh38
Transcripts
NM_001308370.1 NM_194312.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5479567 c.1214A>C p.E405A Chr.2: 238127733-238127733 on Build GRCh38
1018514 c.1151C>A p.P384H Chr.2: 238127670-238127670 on Build GRCh38
4616570 c.1193C>G p.T398S Chr.2: 238127712-238127712 on Build GRCh38
6656362 c.1203G>A p.T401T Chr.2: 238127722-238127722 on Build GRCh38
3748122 c.1120G>C p.A374P Chr.2: 238127639-238127639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750609071 Chr.2: 238127638-238127638 on Build GRCh38
rs753609167 Chr.2: 238127652-238127652 on Build GRCh38
rs62197371 Chr.2: 238127782-238127782 on Build GRCh38
rs998490681 Chr.2: 238127805-238127805 on Build GRCh38
rs141605283 Chr.2: 238127807-238127807 on Build GRCh38
rs986539648 Chr.2: 238127769-238127769 on Build GRCh38
rs748648085 Chr.2: 238127691-238127691 on Build GRCh38
rs963635586 Chr.2: 238127637-238127637 on Build GRCh38
rs774895240 Chr.2: 238127700-238127700 on Build GRCh38
rs777440984 Chr.2: 238127770-238127770 on Build GRCh38
rs747670060 Chr.2: 238127674-238127674 on Build GRCh38
rs763386397 Chr.2: 238127615-238127615 on Build GRCh38
rs761917133 Chr.2: 238127701-238127701 on Build GRCh38
rs773225265 Chr.2: 238127713-238127713 on Build GRCh38
rs765127922 Chr.2: 238127765-238127765 on Build GRCh38
rs754788856 Chr.2: 238127746-238127746 on Build GRCh38
rs12692214 Chr.2: 238127775-238127775 on Build GRCh38
rs752713841 Chr.2: 238127766-238127766 on Build GRCh38
rs766181578 Chr.2: 238127651-238127651 on Build GRCh38
rs200761727 Chr.2: 238127690-238127690 on Build GRCh38
rs760898869 Chr.2: 238127738-238127738 on Build GRCh38
rs1008597127 Chr.2: 238127722-238127722 on Build GRCh38
rs13382446 Chr.2: 238127629-238127629 on Build GRCh38
rs769029179 Chr.2: 238127786-238127786 on Build GRCh38
rs757950744 Chr.2: 238127683-238127683 on Build GRCh38
rs778905030 Chr.2: 238127672-238127672 on Build GRCh38
rs767772256 Chr.2: 238127630-238127630 on Build GRCh38
rs957579501 Chr.2: 238127819-238127819 on Build GRCh38
rs754875593 Chr.2: 238127658-238127658 on Build GRCh38
rs868110740 Chr.2: 238127649-238127649 on Build GRCh38
rs899043573 Chr.2: 238127655-238127655 on Build GRCh38
rs377184238 Chr.2: 238127730-238127730 on Build GRCh38
rs755747094 Chr.2: 238127634-238127634 on Build GRCh38
rs372498854 Chr.2: 238127739-238127739 on Build GRCh38
rs774365332 Chr.2: 238127627-238127627 on Build GRCh38
rs375739536 Chr.2: 238127754-238127754 on Build GRCh38
rs775725938 Chr.2: 238127692-238127692 on Build GRCh38
rs749668238 Chr.2: 238127784-238127784 on Build GRCh38
rs757114674 Chr.2: 238127770-238127771 on Build GRCh38
rs745860787 Chr.2: 238127703-238127703 on Build GRCh38
rs777442317 Chr.2: 238127637-238127638 on Build GRCh38
rs746456370 Chr.2: 238127771-238127771 on Build GRCh38
rs764598149 Chr.2: 238127619-238127619 on Build GRCh38
rs994733458 Chr.2: 238127660-238127660 on Build GRCh38
rs749025781 Chr.2: 238127705-238127707 on Build GRCh38
rs533459489 Chr.2: 238127832-238127832 on Build GRCh38
rs756286557 Chr.2: 238127647-238127647 on Build GRCh38
rs983601951 Chr.2: 238127621-238127621 on Build GRCh38
rs769105149 Chr.2: 238127694-238127694 on Build GRCh38

View Full Product Details