Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 52757798-52758059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTA2
Entrez Gene ID
2939
Gene Name
glutathione S-transferase alpha 2
Gene Aliases
GST2, GSTA2-2, GTA2, GTH2
UniGene
Hs.94107
Gene Chromosome Location
Chr.6: 52750087-52763563 on Build GRCh38
Transcripts
NM_000846.4 XM_011514532.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6309119 c.40G>C p.G14R Chr.6: 52757908-52757908 on Build GRCh38
3949334 c.29C>G p.S10C Chr.6: 52757919-52757919 on Build GRCh38
4586840 c.13C>G p.P5A Chr.6: 52757935-52757935 on Build GRCh38
7126995 c.80G>C p.G27A Chr.6: 52757868-52757868 on Build GRCh38
6577439 c.9G>A p.E3E Chr.6: 52757939-52757939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752329085 Chr.6: 52757823-52757823 on Build GRCh38
rs553440965 Chr.6: 52758025-52758025 on Build GRCh38
rs775912911 Chr.6: 52757845-52757845 on Build GRCh38
rs753628476 Chr.6: 52757878-52757878 on Build GRCh38
rs769933830 Chr.6: 52757972-52757972 on Build GRCh38
rs189908098 Chr.6: 52757863-52757863 on Build GRCh38
rs568358018 Chr.6: 52757861-52757861 on Build GRCh38
rs777437972 Chr.6: 52757825-52757825 on Build GRCh38
rs748524454 Chr.6: 52757907-52757907 on Build GRCh38
rs368366053 Chr.6: 52757831-52757831 on Build GRCh38
rs764298964 Chr.6: 52757983-52757986 on Build GRCh38
rs149555616 Chr.6: 52758015-52758015 on Build GRCh38
rs780064985 Chr.6: 52757955-52757955 on Build GRCh38
rs745987342 Chr.6: 52757833-52757833 on Build GRCh38
rs773702055 Chr.6: 52757857-52757857 on Build GRCh38
rs752078209 Chr.6: 52757983-52757990 on Build GRCh38
rs750201370 Chr.6: 52757935-52757935 on Build GRCh38
rs781442459 Chr.6: 52757903-52757903 on Build GRCh38
rs2254050 Chr.6: 52758016-52758016 on Build GRCh38
rs370331759 Chr.6: 52757871-52757871 on Build GRCh38
rs771481534 Chr.6: 52757988-52757988 on Build GRCh38
rs760170735 Chr.6: 52757995-52757995 on Build GRCh38
rs759542277 Chr.6: 52757984-52757984 on Build GRCh38
rs369593642 Chr.6: 52757856-52757856 on Build GRCh38
rs201934364 Chr.6: 52757868-52757868 on Build GRCh38
rs770775412 Chr.6: 52757925-52757925 on Build GRCh38
rs184931824 Chr.6: 52757982-52757982 on Build GRCh38
rs183965512 Chr.6: 52758017-52758017 on Build GRCh38
rs146322588 Chr.6: 52757909-52757909 on Build GRCh38
rs200631253 Chr.6: 52757930-52757930 on Build GRCh38
rs752975446 Chr.6: 52757979-52757986 on Build GRCh38
rs779423039 Chr.6: 52757859-52757859 on Build GRCh38
rs201154224 Chr.6: 52757867-52757867 on Build GRCh38
rs768883687 Chr.6: 52757849-52757849 on Build GRCh38
rs201315214 Chr.6: 52757882-52757882 on Build GRCh38
rs776880388 Chr.6: 52757850-52757850 on Build GRCh38
rs758496844 Chr.6: 52757982-52757983 on Build GRCh38
rs150418183 Chr.6: 52757991-52757991 on Build GRCh38
rs781603944 Chr.6: 52757993-52757993 on Build GRCh38
rs142063997 Chr.6: 52757889-52757889 on Build GRCh38
rs541904401 Chr.6: 52757910-52757910 on Build GRCh38
rs747060432 Chr.6: 52757892-52757892 on Build GRCh38
rs375054470 Chr.6: 52757824-52757824 on Build GRCh38
rs772274379 Chr.6: 52757844-52757844 on Build GRCh38
rs139552194 Chr.6: 52757926-52757926 on Build GRCh38
rs751630345 Chr.6: 52757953-52757953 on Build GRCh38
rs113239409 Chr.6: 52757864-52757864 on Build GRCh38
rs762215120 Chr.6: 52757852-52757852 on Build GRCh38
rs748413219 Chr.6: 52757958-52757958 on Build GRCh38
rs771389883 Chr.6: 52757911-52757911 on Build GRCh38
rs386701157 Chr.6: 52758012-52758021 on Build GRCh38
rs747200068 Chr.6: 52757847-52757847 on Build GRCh38
rs749607946 Chr.6: 52757986-52757986 on Build GRCh38
rs1803684 Chr.6: 52757957-52757957 on Build GRCh38
rs568198841 Chr.6: 52758020-52758020 on Build GRCh38
rs780311849 Chr.6: 52757891-52757891 on Build GRCh38
rs909873779 Chr.6: 52758019-52758019 on Build GRCh38
rs758343016 Chr.6: 52757890-52757890 on Build GRCh38
rs780117604 Chr.6: 52757949-52757949 on Build GRCh38
rs373318601 Chr.6: 52757921-52757921 on Build GRCh38
rs533894957 Chr.6: 52758021-52758021 on Build GRCh38
rs778973127 Chr.6: 52757832-52757832 on Build GRCh38
rs377509381 Chr.6: 52757924-52757924 on Build GRCh38
rs757047103 Chr.6: 52757881-52757881 on Build GRCh38
rs138220898 Chr.6: 52758008-52758008 on Build GRCh38
rs757750531 Chr.6: 52757990-52757991 on Build GRCh38
rs190967828 Chr.6: 52758012-52758012 on Build GRCh38
rs758431120 Chr.6: 52757947-52757947 on Build GRCh38
rs376966272 Chr.6: 52757963-52757963 on Build GRCh38

View Full Product Details