Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44143146-44143419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf33
Entrez Gene ID
8209
Gene Name
chromosome 21 open reading frame 33
Gene Aliases
ES1, GT335, HES1, KNPH, KNPI
UniGene
Hs.413482
Gene Chromosome Location
Chr.21: 44133612-44145723 on Build GRCh38
Transcripts
NM_004649.7 NM_198155.4 NR_135220.1 XM_017028479.1 XM_006723901.3 XM_006723903.3 XM_011546081.2 XM_017028514.1 XM_017028516.1 XM_017028517.1

Gene Symbol
LOC102724023
Entrez Gene ID
102724023
Gene Name
ES1 protein homolog, mitochondrial
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.21: 5097172-5128463 on Build GRCh38
Transcripts
NM_004649.7 NM_198155.4 NR_135220.1 XM_017028479.1 XM_006723901.3 XM_006723903.3 XM_011546081.2 XM_017028514.1 XM_017028516.1 XM_017028517.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2820513 c.664G>A p.V222M Chr.21: 44143347-44143347 on Build GRCh38
7149088 c.631G>C p.E211Q Chr.21: 44143314-44143314 on Build GRCh38
7116734 c.605G>A p.G202D Chr.21: 44143288-44143288 on Build GRCh38
6396711 c.609G>T p.K203N Chr.21: 44143292-44143292 on Build GRCh38
231900 c.598G>A p.E200K Chr.21: 44143281-44143281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112869363 Chr.21: 44143362-44143362 on Build GRCh38
rs760714300 Chr.21: 44143304-44143304 on Build GRCh38
rs752956051 Chr.21: 44143322-44143322 on Build GRCh38
rs747323775 Chr.21: 44143339-44143339 on Build GRCh38
rs759604742 Chr.21: 44143390-44143390 on Build GRCh38
rs747811900 Chr.21: 44143360-44143360 on Build GRCh38
rs753923011 Chr.21: 44143238-44143238 on Build GRCh38
rs767871364 Chr.21: 44143351-44143352 on Build GRCh38
rs149515079 Chr.21: 44143305-44143305 on Build GRCh38
rs769116753 Chr.21: 44143176-44143176 on Build GRCh38
rs775001279 Chr.21: 44143273-44143273 on Build GRCh38
rs772465919 Chr.21: 44143254-44143254 on Build GRCh38
rs752032193 Chr.21: 44143284-44143284 on Build GRCh38
rs574424318 Chr.21: 44143302-44143302 on Build GRCh38
rs376199934 Chr.21: 44143396-44143396 on Build GRCh38
rs751341818 Chr.21: 44143221-44143221 on Build GRCh38
rs777661334 Chr.21: 44143386-44143386 on Build GRCh38
rs781612751 Chr.21: 44143346-44143346 on Build GRCh38
rs148646039 Chr.21: 44143324-44143324 on Build GRCh38
rs200229685 Chr.21: 44143180-44143180 on Build GRCh38
rs749492834 Chr.21: 44143171-44143171 on Build GRCh38
rs143937665 Chr.21: 44143310-44143310 on Build GRCh38
rs779170823 Chr.21: 44143184-44143184 on Build GRCh38
rs369189366 Chr.21: 44143397-44143397 on Build GRCh38
rs201483651 Chr.21: 44143320-44143320 on Build GRCh38
rs531379635 Chr.21: 44143347-44143347 on Build GRCh38
rs372877054 Chr.21: 44143299-44143299 on Build GRCh38
rs779128079 Chr.21: 44143251-44143251 on Build GRCh38
rs144188289 Chr.21: 44143361-44143361 on Build GRCh38
rs761610960 Chr.21: 44143229-44143229 on Build GRCh38
rs149440055 Chr.21: 44143185-44143185 on Build GRCh38
rs748471304 Chr.21: 44143253-44143253 on Build GRCh38
rs372989488 Chr.21: 44143391-44143391 on Build GRCh38
rs146274470 Chr.21: 44143232-44143232 on Build GRCh38
rs769341133 Chr.21: 44143272-44143272 on Build GRCh38
rs752914470 Chr.21: 44143394-44143394 on Build GRCh38
rs199604151 Chr.21: 44143271-44143271 on Build GRCh38
rs762484928 Chr.21: 44143278-44143278 on Build GRCh38
rs756303766 Chr.21: 44143231-44143231 on Build GRCh38
rs755189625 Chr.21: 44143239-44143239 on Build GRCh38
rs139126084 Chr.21: 44143263-44143263 on Build GRCh38
rs112995189 Chr.21: 44143363-44143363 on Build GRCh38
rs750436429 Chr.21: 44143303-44143303 on Build GRCh38
rs762489987 Chr.21: 44143198-44143198 on Build GRCh38
rs768197728 Chr.21: 44143279-44143279 on Build GRCh38
rs200252119 Chr.21: 44143196-44143196 on Build GRCh38
rs192398836 Chr.21: 44143230-44143230 on Build GRCh38
rs760476789 Chr.21: 44143370-44143370 on Build GRCh38
rs778086540 Chr.21: 44143325-44143325 on Build GRCh38
rs533182277 Chr.21: 44143311-44143311 on Build GRCh38
rs994776203 Chr.21: 44143220-44143220 on Build GRCh38
rs763752092 Chr.21: 44143207-44143207 on Build GRCh38
rs548701197 Chr.21: 44143346-44143347 on Build GRCh38
rs763322397 Chr.21: 44143237-44143254 on Build GRCh38
rs757790799 Chr.21: 44143342-44143342 on Build GRCh38
rs764357327 Chr.21: 44143246-44143253 on Build GRCh38
rs370573156 Chr.21: 44143385-44143385 on Build GRCh38
rs773913787 Chr.21: 44143348-44143348 on Build GRCh38
rs754540861 Chr.21: 44143195-44143195 on Build GRCh38
rs201295411 Chr.21: 44143371-44143371 on Build GRCh38

View Full Product Details