Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37303225-37303458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH4
Entrez Gene ID
27128
Gene Name
cytohesin 4
Gene Aliases
CYT4, DJ63G5.1, PSCD4
UniGene
Hs.170944
Gene Chromosome Location
Chr.22: 37282383-37315348 on Build GRCh38
Transcripts
NM_001318024.1 NM_013385.4 XM_011530147.2 XM_011530148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7196387 c.663C>A p.N221K Chr.22: 37303369-37303369 on Build GRCh38
444958 c.683A>G p.E228G Chr.22: 37303389-37303389 on Build GRCh38
5773870 c.675C>T p.D225D Chr.22: 37303381-37303381 on Build GRCh38
3150301 c.559G>A p.V187M Chr.22: 37303265-37303265 on Build GRCh38
6639606 c.694C>T p.R232W Chr.22: 37303400-37303400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760831589 Chr.22: 37303425-37303425 on Build GRCh38
rs374106882 Chr.22: 37303250-37303250 on Build GRCh38
rs746552896 Chr.22: 37303340-37303340 on Build GRCh38
rs765068937 Chr.22: 37303381-37303381 on Build GRCh38
rs184409484 Chr.22: 37303360-37303360 on Build GRCh38
rs377543427 Chr.22: 37303356-37303356 on Build GRCh38
rs375468773 Chr.22: 37303400-37303400 on Build GRCh38
rs371808106 Chr.22: 37303399-37303399 on Build GRCh38
rs369723358 Chr.22: 37303249-37303249 on Build GRCh38
rs766400498 Chr.22: 37303375-37303375 on Build GRCh38
rs989547217 Chr.22: 37303246-37303246 on Build GRCh38
rs148578667 Chr.22: 37303264-37303264 on Build GRCh38
rs141320333 Chr.22: 37303387-37303387 on Build GRCh38
rs371162850 Chr.22: 37303358-37303358 on Build GRCh38
rs962420964 Chr.22: 37303329-37303329 on Build GRCh38
rs753735770 Chr.22: 37303377-37303377 on Build GRCh38
rs2239822 Chr.22: 37303336-37303336 on Build GRCh38
rs773391601 Chr.22: 37303367-37303367 on Build GRCh38
rs754755137 Chr.22: 37303327-37303327 on Build GRCh38
rs373433486 Chr.22: 37303328-37303328 on Build GRCh38
rs764469352 Chr.22: 37303283-37303283 on Build GRCh38
rs774848140 Chr.22: 37303359-37303359 on Build GRCh38
rs748552711 Chr.22: 37303413-37303413 on Build GRCh38
rs747888441 Chr.22: 37303330-37303330 on Build GRCh38
rs202039940 Chr.22: 37303378-37303378 on Build GRCh38
rs982103198 Chr.22: 37303270-37303270 on Build GRCh38
rs756164286 Chr.22: 37303306-37303306 on Build GRCh38
rs200267703 Chr.22: 37303401-37303401 on Build GRCh38
rs750560096 Chr.22: 37303295-37303295 on Build GRCh38
rs776827869 Chr.22: 37303431-37303431 on Build GRCh38
rs764794103 Chr.22: 37303421-37303422 on Build GRCh38
rs534559418 Chr.22: 37303393-37303393 on Build GRCh38
rs753773355 Chr.22: 37303320-37303320 on Build GRCh38
rs146595470 Chr.22: 37303346-37303346 on Build GRCh38
rs374132944 Chr.22: 37303411-37303411 on Build GRCh38
rs116494108 Chr.22: 37303435-37303435 on Build GRCh38
rs760778626 Chr.22: 37303374-37303374 on Build GRCh38
rs767776599 Chr.22: 37303361-37303361 on Build GRCh38
rs375934892 Chr.22: 37303436-37303436 on Build GRCh38
rs201940219 Chr.22: 37303388-37303388 on Build GRCh38
rs777541367 Chr.22: 37303337-37303337 on Build GRCh38
rs767723450 Chr.22: 37303286-37303286 on Build GRCh38
rs771005479 Chr.22: 37303429-37303429 on Build GRCh38
rs756120570 Chr.22: 37303265-37303265 on Build GRCh38
rs553131134 Chr.22: 37303414-37303414 on Build GRCh38
rs769265920 Chr.22: 37303409-37303409 on Build GRCh38
rs774447340 Chr.22: 37303284-37303284 on Build GRCh38
rs766349067 Chr.22: 37303319-37303319 on Build GRCh38
rs776109061 Chr.22: 37303273-37303273 on Build GRCh38
rs773448811 Chr.22: 37303424-37303424 on Build GRCh38
rs373821857 Chr.22: 37303379-37303379 on Build GRCh38
rs202195060 Chr.22: 37303282-37303282 on Build GRCh38
rs770524744 Chr.22: 37303341-37303341 on Build GRCh38
rs762030009 Chr.22: 37303285-37303285 on Build GRCh38
rs35078298 Chr.22: 37303387-37303388 on Build GRCh38

View Full Product Details