Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44550773-44551024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3J
Entrez Gene ID
8669
Gene Name
eukaryotic translation initiation factor 3 subunit J
Gene Aliases
EIF3S1, eIF3-alpha, eIF3-p35
UniGene
Hs.404056
Gene Chromosome Location
Chr.15: 44537068-44562803 on Build GRCh38
Transcripts
NM_001284335.1 NM_001284336.1 NM_003758.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652866 c.162C>T p.D54D Chr.15: 44550890-44550890 on Build GRCh38
3686100 c.191_194delAAGT p.V65fs*4 Chr.15: 44550919-44550922 on Build GRCh38
1373100 c.173delA p.E61fs*5 Chr.15: 44550901-44550901 on Build GRCh38
291018 c.173_174delAA p.K60fs*36 Chr.15: 44550901-44550902 on Build GRCh38
962104 c.187G>A p.A63T Chr.15: 44550915-44550915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755095039 Chr.15: 44550870-44550870 on Build GRCh38
rs147844807 Chr.15: 44550945-44550945 on Build GRCh38
rs974681607 Chr.15: 44550813-44550813 on Build GRCh38
rs1016712152 Chr.15: 44550946-44550946 on Build GRCh38
rs895087016 Chr.15: 44550985-44550985 on Build GRCh38
rs770760665 Chr.15: 44550898-44550898 on Build GRCh38
rs1029902423 Chr.15: 44550992-44550992 on Build GRCh38
rs775994053 Chr.15: 44550921-44550921 on Build GRCh38
rs751658101 Chr.15: 44550858-44550858 on Build GRCh38
rs367949712 Chr.15: 44550966-44550966 on Build GRCh38
rs975309909 Chr.15: 44550991-44550991 on Build GRCh38
rs750697496 Chr.15: 44550828-44550828 on Build GRCh38
rs1055046994 Chr.15: 44550984-44550984 on Build GRCh38
rs768349793 Chr.15: 44550918-44550918 on Build GRCh38
rs765601047 Chr.15: 44550948-44550948 on Build GRCh38
rs745581861 Chr.15: 44550900-44550900 on Build GRCh38
rs141093779 Chr.15: 44550909-44550909 on Build GRCh38
rs758626231 Chr.15: 44550842-44550842 on Build GRCh38
rs113111320 Chr.15: 44550931-44550931 on Build GRCh38
rs549570967 Chr.15: 44550811-44550811 on Build GRCh38
rs930071579 Chr.15: 44550905-44550905 on Build GRCh38
rs937339817 Chr.15: 44550983-44550983 on Build GRCh38
rs577541826 Chr.15: 44550804-44550804 on Build GRCh38
rs146638481 Chr.15: 44550890-44550890 on Build GRCh38
rs760374692 Chr.15: 44550911-44550911 on Build GRCh38
rs778780412 Chr.15: 44550899-44550899 on Build GRCh38
rs368372196 Chr.15: 44550876-44550876 on Build GRCh38
rs749313160 Chr.15: 44550893-44550893 on Build GRCh38
rs780205818 Chr.15: 44550857-44550857 on Build GRCh38
rs200969499 Chr.15: 44550923-44550923 on Build GRCh38
rs750834049 Chr.15: 44550953-44550953 on Build GRCh38
rs771873931 Chr.15: 44550901-44550901 on Build GRCh38
rs756653669 Chr.15: 44550891-44550893 on Build GRCh38
rs755511421 Chr.15: 44550909-44550911 on Build GRCh38

View Full Product Details