Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77878918-77879171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAMDC
Entrez Gene ID
28971
Gene Name
adipogenesis associated Mth938 domain containing
Gene Aliases
C11orf67, CK067, PTD015
UniGene
Hs.745040
Gene Chromosome Location
Chr.11: 77821114-77901281 on Build GRCh38
Transcripts
NM_001316957.1 NM_001316958.1 NR_133637.1 XM_017017620.1 NM_033547.3 XM_011545352.2 XM_017018560.1

Gene Symbol
INTS4
Entrez Gene ID
92105
Gene Name
integrator complex subunit 4
Gene Aliases
INT4, MST093
UniGene
Hs.533723
Gene Chromosome Location
Chr.11: 77874491-77994849 on Build GRCh38
Transcripts
NM_001316957.1 NM_001316958.1 NR_133637.1 XM_017017620.1 NM_033547.3 XM_011545352.2 XM_017018560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4146353 c.2796T>G p.G932G Chr.11: 77879045-77879045 on Build GRCh38
4757800 c.2765G>A p.R922H Chr.11: 77879076-77879076 on Build GRCh38
6686456 c.2746G>A p.A916T Chr.11: 77879095-77879095 on Build GRCh38
458369 c.2826C>T p.I942I Chr.11: 77879015-77879015 on Build GRCh38
5006741 c.2887C>T p.R963C Chr.11: 77878954-77878954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776901103 Chr.11: 77878951-77878951 on Build GRCh38
rs761947815 Chr.11: 77879090-77879090 on Build GRCh38
rs769873160 Chr.11: 77879097-77879098 on Build GRCh38
rs771692626 Chr.11: 77879027-77879027 on Build GRCh38
rs746252330 Chr.11: 77879039-77879039 on Build GRCh38
rs777902160 Chr.11: 77879130-77879131 on Build GRCh38
rs747665040 Chr.11: 77879024-77879024 on Build GRCh38
rs751824347 Chr.11: 77878962-77878962 on Build GRCh38
rs200717599 Chr.11: 77879135-77879135 on Build GRCh38
rs748853100 Chr.11: 77878938-77878938 on Build GRCh38
rs367591609 Chr.11: 77879019-77879019 on Build GRCh38
rs774332448 Chr.11: 77879086-77879086 on Build GRCh38
rs757869752 Chr.11: 77879140-77879140 on Build GRCh38
rs141434365 Chr.11: 77879029-77879029 on Build GRCh38
rs777576154 Chr.11: 77879143-77879143 on Build GRCh38
rs140409215 Chr.11: 77878971-77878971 on Build GRCh38
rs764036133 Chr.11: 77878961-77878961 on Build GRCh38
rs760467454 Chr.11: 77879098-77879098 on Build GRCh38
rs185402665 Chr.11: 77878948-77878948 on Build GRCh38
rs201190994 Chr.11: 77879145-77879145 on Build GRCh38
rs778338283 Chr.11: 77879022-77879022 on Build GRCh38
rs754539518 Chr.11: 77879021-77879021 on Build GRCh38
rs188994465 Chr.11: 77878990-77878990 on Build GRCh38
rs757466320 Chr.11: 77878963-77878963 on Build GRCh38
rs769182209 Chr.11: 77879077-77879077 on Build GRCh38
rs749185225 Chr.11: 77878994-77878994 on Build GRCh38
rs536814853 Chr.11: 77878956-77878956 on Build GRCh38
rs770220271 Chr.11: 77879062-77879062 on Build GRCh38
rs753607740 Chr.11: 77879122-77879122 on Build GRCh38
rs754914473 Chr.11: 77879134-77879134 on Build GRCh38
rs765904872 Chr.11: 77878954-77878954 on Build GRCh38
rs543195366 Chr.11: 77879104-77879104 on Build GRCh38
rs763501825 Chr.11: 77879076-77879076 on Build GRCh38
rs747706522 Chr.11: 77878943-77878943 on Build GRCh38
rs759978044 Chr.11: 77878953-77878953 on Build GRCh38
rs768095858 Chr.11: 77878939-77878939 on Build GRCh38
rs948926154 Chr.11: 77879094-77879094 on Build GRCh38
rs1054909908 Chr.11: 77879085-77879085 on Build GRCh38
rs774085027 Chr.11: 77878940-77878940 on Build GRCh38
rs779979726 Chr.11: 77878991-77878991 on Build GRCh38
rs374670901 Chr.11: 77878957-77878957 on Build GRCh38
rs750284357 Chr.11: 77878984-77878984 on Build GRCh38
rs772908378 Chr.11: 77879035-77879035 on Build GRCh38
rs750586409 Chr.11: 77879096-77879096 on Build GRCh38
rs752209550 Chr.11: 77879138-77879138 on Build GRCh38
rs370175361 Chr.11: 77879101-77879101 on Build GRCh38
rs1046744383 Chr.11: 77878967-77878967 on Build GRCh38

View Full Product Details