Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67289478-67289749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD19
Entrez Gene ID
146212
Gene Name
potassium channel tetramerization domain containing 19
Gene Aliases
-
UniGene
Hs.299127
Gene Chromosome Location
Chr.16: 67289490-67326758 on Build GRCh38
Transcripts
NM_001100915.1 NM_001129727.2 NM_001129728.1 NM_001129729.2 NM_001129731.2 XM_011522985.2 XM_011522986.2 XM_011522987.2 XM_011522988.2

Gene Symbol
PLEKHG4
Entrez Gene ID
25894
Gene Name
pleckstrin homology and RhoGEF domain containing G4
Gene Aliases
ARHGEF44, PRTPHN1, SCA4
UniGene
Hs.188781
Gene Chromosome Location
Chr.16: 67277820-67289500 on Build GRCh38
Transcripts
NM_001100915.1 NM_001129727.2 NM_001129728.1 NM_001129729.2 NM_001129731.2 XM_011522985.2 XM_011522986.2 XM_011522987.2 XM_011522988.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4694810 c.2739T>C p.S913S Chr.16: 67289611-67289611 on Build GRCh38
4061830 c.2669T>G p.L890R Chr.16: 67289681-67289681 on Build GRCh38
4579158 c.2674C>T p.L892L Chr.16: 67289676-67289676 on Build GRCh38
3511003 c.2735G>A p.R912K Chr.16: 67289615-67289615 on Build GRCh38
4061829 c.2679C>A p.P893P Chr.16: 67289671-67289671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017457136 Chr.16: 67289708-67289708 on Build GRCh38
rs528509208 Chr.16: 67289605-67289605 on Build GRCh38
rs765090605 Chr.16: 67289666-67289666 on Build GRCh38
rs751192202 Chr.16: 67289672-67289672 on Build GRCh38
rs760432260 Chr.16: 67289626-67289626 on Build GRCh38
rs202058376 Chr.16: 67289676-67289676 on Build GRCh38
rs774712571 Chr.16: 67289625-67289625 on Build GRCh38
rs887845042 Chr.16: 67289569-67289569 on Build GRCh38
rs571668629 Chr.16: 67289695-67289695 on Build GRCh38
rs1028282262 Chr.16: 67289546-67289546 on Build GRCh38
rs780696174 Chr.16: 67289689-67289689 on Build GRCh38
rs752161156 Chr.16: 67289555-67289555 on Build GRCh38
rs781644473 Chr.16: 67289678-67289678 on Build GRCh38
rs766298598 Chr.16: 67289538-67289538 on Build GRCh38
rs777468272 Chr.16: 67289671-67289671 on Build GRCh38
rs766170854 Chr.16: 67289638-67289638 on Build GRCh38
rs775718313 Chr.16: 67289600-67289600 on Build GRCh38
rs369273590 Chr.16: 67289596-67289596 on Build GRCh38
rs763301444 Chr.16: 67289529-67289529 on Build GRCh38
rs1013797000 Chr.16: 67289620-67289620 on Build GRCh38
rs777348066 Chr.16: 67289577-67289577 on Build GRCh38
rs371085149 Chr.16: 67289668-67289668 on Build GRCh38
rs997986285 Chr.16: 67289510-67289510 on Build GRCh38
rs201253917 Chr.16: 67289652-67289652 on Build GRCh38
rs374179662 Chr.16: 67289545-67289545 on Build GRCh38
rs748421148 Chr.16: 67289702-67289702 on Build GRCh38
rs747389979 Chr.16: 67289586-67289586 on Build GRCh38
rs756704258 Chr.16: 67289684-67289684 on Build GRCh38
rs370542549 Chr.16: 67289706-67289706 on Build GRCh38
rs781589008 Chr.16: 67289588-67289588 on Build GRCh38
rs746409848 Chr.16: 67289592-67289592 on Build GRCh38
rs370642604 Chr.16: 67289697-67289697 on Build GRCh38
rs758048004 Chr.16: 67289558-67289558 on Build GRCh38
rs754491719 Chr.16: 67289543-67289543 on Build GRCh38
rs200200525 Chr.16: 67289645-67289645 on Build GRCh38
rs771321349 Chr.16: 67289587-67289587 on Build GRCh38
rs753786814 Chr.16: 67289539-67289539 on Build GRCh38
rs930886394 Chr.16: 67289714-67289714 on Build GRCh38
rs369315078 Chr.16: 67289535-67289535 on Build GRCh38
rs376980368 Chr.16: 67289680-67289680 on Build GRCh38
rs759546737 Chr.16: 67289656-67289656 on Build GRCh38
rs561137674 Chr.16: 67289707-67289707 on Build GRCh38
rs547035454 Chr.16: 67289623-67289623 on Build GRCh38
rs374336918 Chr.16: 67289704-67289704 on Build GRCh38
rs1047830551 Chr.16: 67289567-67289567 on Build GRCh38
rs373147643 Chr.16: 67289599-67289599 on Build GRCh38
rs752214690 Chr.16: 67289667-67289667 on Build GRCh38
rs764346258 Chr.16: 67289534-67289534 on Build GRCh38
rs377068044 Chr.16: 67289641-67289641 on Build GRCh38

View Full Product Details