Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 68046694-68046967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIRAS3
Entrez Gene ID
9077
Gene Name
DIRAS family GTPase 3
Gene Aliases
ARHI, NOEY2
UniGene
Hs.194695
Gene Chromosome Location
Chr.1: 68045962-68051762 on Build GRCh38
Transcripts
NM_004675.3 XM_017002773.1 NR_040077.1

Gene Symbol
GNG12-AS1
Entrez Gene ID
100289178
Gene Name
GNG12 antisense RNA 1
Gene Aliases
-
UniGene
Hs.677603
Gene Chromosome Location
Chr.1: 67832288-68202987 on Build GRCh38
Transcripts
NM_004675.3 XM_017002773.1 NR_040077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
911851 c.386C>T p.A129V Chr.1: 68046912-68046912 on Build GRCh38
5630451 c.362A>G p.K121R Chr.1: 68046936-68046936 on Build GRCh38
4009674 c.551C>G p.T184S Chr.1: 68046747-68046747 on Build GRCh38
5590531 c.480G>A p.E160E Chr.1: 68046818-68046818 on Build GRCh38
911850 c.402C>A p.I134I Chr.1: 68046896-68046896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761694570 Chr.1: 68046856-68046856 on Build GRCh38
rs1049128443 Chr.1: 68046728-68046728 on Build GRCh38
rs779753508 Chr.1: 68046800-68046800 on Build GRCh38
rs751418374 Chr.1: 68046778-68046778 on Build GRCh38
rs778723432 Chr.1: 68046913-68046913 on Build GRCh38
rs776658969 Chr.1: 68046734-68046734 on Build GRCh38
rs759605194 Chr.1: 68046845-68046845 on Build GRCh38
rs755683723 Chr.1: 68046795-68046795 on Build GRCh38
rs750283758 Chr.1: 68046792-68046792 on Build GRCh38
rs771445902 Chr.1: 68046827-68046827 on Build GRCh38
rs758037524 Chr.1: 68046917-68046917 on Build GRCh38
rs61736596 Chr.1: 68046758-68046758 on Build GRCh38
rs778375581 Chr.1: 68046824-68046824 on Build GRCh38
rs375502625 Chr.1: 68046801-68046801 on Build GRCh38
rs778048012 Chr.1: 68046942-68046942 on Build GRCh38
rs201724675 Chr.1: 68046810-68046810 on Build GRCh38
rs768461188 Chr.1: 68046859-68046860 on Build GRCh38
rs930742459 Chr.1: 68046721-68046721 on Build GRCh38
rs767507651 Chr.1: 68046788-68046788 on Build GRCh38
rs898799980 Chr.1: 68046756-68046756 on Build GRCh38
rs370918535 Chr.1: 68046772-68046772 on Build GRCh38
rs368845838 Chr.1: 68046870-68046870 on Build GRCh38
rs776746697 Chr.1: 68046830-68046830 on Build GRCh38
rs766407363 Chr.1: 68046874-68046874 on Build GRCh38
rs147087321 Chr.1: 68046904-68046904 on Build GRCh38
rs754473486 Chr.1: 68046907-68046907 on Build GRCh38
rs987306069 Chr.1: 68046813-68046813 on Build GRCh38
rs748551952 Chr.1: 68046822-68046822 on Build GRCh38
rs775628945 Chr.1: 68046854-68046854 on Build GRCh38
rs553888339 Chr.1: 68046857-68046857 on Build GRCh38
rs766327434 Chr.1: 68046725-68046725 on Build GRCh38
rs149791622 Chr.1: 68046809-68046809 on Build GRCh38
rs1031312142 Chr.1: 68046872-68046872 on Build GRCh38
rs747508747 Chr.1: 68046825-68046825 on Build GRCh38
rs770003646 Chr.1: 68046849-68046849 on Build GRCh38
rs753465480 Chr.1: 68046882-68046882 on Build GRCh38
rs894676240 Chr.1: 68046733-68046733 on Build GRCh38
rs761443919 Chr.1: 68046785-68046785 on Build GRCh38
rs542823739 Chr.1: 68046823-68046823 on Build GRCh38
rs964544940 Chr.1: 68046881-68046881 on Build GRCh38

View Full Product Details