Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2717100-2717327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIRAS1
Entrez Gene ID
148252
Gene Name
DIRAS family GTPase 1
Gene Aliases
Di-Ras1, GBTS1, RIG
UniGene
Hs.172753
Gene Chromosome Location
Chr.19: 2714567-2721392 on Build GRCh38
Transcripts
NM_145173.3 XM_011527713.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2749539 c.547G>A p.G183R Chr.19: 2717260-2717260 on Build GRCh38
6403492 c.497C>T p.T166M Chr.19: 2717310-2717310 on Build GRCh38
6400277 c.583T>C p.C195R Chr.19: 2717224-2717224 on Build GRCh38
6644226 c.532G>A p.D178N Chr.19: 2717275-2717275 on Build GRCh38
378251 c.545C>T p.S182F Chr.19: 2717262-2717262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113069103 Chr.19: 2717206-2717206 on Build GRCh38
rs143766646 Chr.19: 2717273-2717273 on Build GRCh38
rs146665806 Chr.19: 2717166-2717166 on Build GRCh38
rs753227238 Chr.19: 2717233-2717234 on Build GRCh38
rs866466185 Chr.19: 2717146-2717146 on Build GRCh38
rs777899917 Chr.19: 2717261-2717261 on Build GRCh38
rs1053012805 Chr.19: 2717135-2717135 on Build GRCh38
rs766301692 Chr.19: 2717286-2717286 on Build GRCh38
rs376093330 Chr.19: 2717183-2717183 on Build GRCh38
rs776453682 Chr.19: 2717201-2717201 on Build GRCh38
rs761356273 Chr.19: 2717205-2717205 on Build GRCh38
rs146622549 Chr.19: 2717255-2717255 on Build GRCh38
rs763042277 Chr.19: 2717214-2717214 on Build GRCh38
rs747806974 Chr.19: 2717279-2717279 on Build GRCh38
rs757827009 Chr.19: 2717272-2717272 on Build GRCh38
rs766065756 Chr.19: 2717222-2717222 on Build GRCh38
rs768391370 Chr.19: 2717197-2717197 on Build GRCh38
rs201931758 Chr.19: 2717288-2717288 on Build GRCh38
rs201111715 Chr.19: 2717180-2717180 on Build GRCh38
rs760219986 Chr.19: 2717185-2717185 on Build GRCh38
rs752915565 Chr.19: 2717295-2717295 on Build GRCh38
rs769887480 Chr.19: 2717169-2717169 on Build GRCh38
rs772343054 Chr.19: 2717275-2717275 on Build GRCh38
rs977431561 Chr.19: 2717138-2717138 on Build GRCh38
rs558633973 Chr.19: 2717285-2717285 on Build GRCh38
rs908236028 Chr.19: 2717188-2717188 on Build GRCh38
rs3745812 Chr.19: 2717258-2717258 on Build GRCh38
rs913104935 Chr.19: 2717142-2717142 on Build GRCh38
rs187199591 Chr.19: 2717144-2717144 on Build GRCh38
rs941104514 Chr.19: 2717221-2717221 on Build GRCh38
rs769416277 Chr.19: 2717282-2717282 on Build GRCh38
rs765912241 Chr.19: 2717171-2717175 on Build GRCh38
rs762269587 Chr.19: 2717160-2717161 on Build GRCh38
rs922896347 Chr.19: 2717143-2717143 on Build GRCh38
rs200400450 Chr.19: 2717265-2717265 on Build GRCh38
rs748189055 Chr.19: 2717167-2717167 on Build GRCh38
rs750317838 Chr.19: 2717211-2717211 on Build GRCh38
rs1037528236 Chr.19: 2717236-2717236 on Build GRCh38
rs751412810 Chr.19: 2717238-2717238 on Build GRCh38
rs767308385 Chr.19: 2717294-2717294 on Build GRCh38
rs1043163856 Chr.19: 2717220-2717220 on Build GRCh38
rs536429882 Chr.19: 2717181-2717181 on Build GRCh38
rs781430267 Chr.19: 2717243-2717243 on Build GRCh38
rs371646644 Chr.19: 2717239-2717239 on Build GRCh38
rs778199016 Chr.19: 2717170-2717170 on Build GRCh38
rs776364121 Chr.19: 2717276-2717276 on Build GRCh38

View Full Product Details