Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 46575290-46575533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST7
Entrez Gene ID
56548
Gene Name
carbohydrate sulfotransferase 7
Gene Aliases
C6ST-2, GST-5
UniGene
Hs.129955
Gene Chromosome Location
Chr.X: 46573687-46598496 on Build GRCh38
Transcripts
NM_019886.3

SNP Detail

SNP ID Chromosome Location
rs771942314 Chr.X: 46575389-46575389 on Build GRCh38
rs763536246 Chr.X: 46575431-46575431 on Build GRCh38
rs771178040 Chr.X: 46575362-46575362 on Build GRCh38
rs908302858 Chr.X: 46575404-46575404 on Build GRCh38
rs754438460 Chr.X: 46575353-46575353 on Build GRCh38
rs748282034 Chr.X: 46575329-46575329 on Build GRCh38
rs760200031 Chr.X: 46575422-46575422 on Build GRCh38
rs780218337 Chr.X: 46575313-46575313 on Build GRCh38
rs1035920889 Chr.X: 46575415-46575415 on Build GRCh38
rs755496206 Chr.X: 46575318-46575318 on Build GRCh38
rs968871297 Chr.X: 46575469-46575469 on Build GRCh38
rs758048006 Chr.X: 46575503-46575503 on Build GRCh38
rs754413283 Chr.X: 46575323-46575323 on Build GRCh38
rs1003140372 Chr.X: 46575349-46575349 on Build GRCh38
rs775365574 Chr.X: 46575416-46575416 on Build GRCh38
rs774067139 Chr.X: 46575368-46575368 on Build GRCh38
rs917159029 Chr.X: 46575452-46575452 on Build GRCh38
rs1021796123 Chr.X: 46575436-46575436 on Build GRCh38
rs140493918 Chr.X: 46575457-46575457 on Build GRCh38
rs778539232 Chr.X: 46575420-46575420 on Build GRCh38
rs961173933 Chr.X: 46575410-46575410 on Build GRCh38
rs773594116 Chr.X: 46575360-46575360 on Build GRCh38
rs749451933 Chr.X: 46575361-46575361 on Build GRCh38
rs769957315 Chr.X: 46575337-46575337 on Build GRCh38
rs374354748 Chr.X: 46575312-46575312 on Build GRCh38
rs983852290 Chr.X: 46575401-46575401 on Build GRCh38
rs368196966 Chr.X: 46575377-46575377 on Build GRCh38
rs866358724 Chr.X: 46575358-46575358 on Build GRCh38
rs747397911 Chr.X: 46575315-46575315 on Build GRCh38

View Full Product Details