Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 8855119-8855362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARHSP1
Entrez Gene ID
23589
Gene Name
calcium regulated heat stable protein 1
Gene Aliases
CRHSP-24, CRHSP24, CSDC1
UniGene
Hs.632184
Gene Chromosome Location
Chr.16: 8852942-8869012 on Build GRCh38
Transcripts
NM_001042476.2 NM_001278260.1 NM_001278261.1 NM_001278262.1 NM_001278263.1 NM_001278264.1 NM_001278265.1 NM_001278266.1 NM_014316.3 XM_005255228.4 XM_005255229.4 XM_011522444.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2923149 c.313G>A p.D105N Chr.16: 8855295-8855295 on Build GRCh38
6225121 c.315C>T p.D105D Chr.16: 8855293-8855293 on Build GRCh38
4669269 c.348delC p.K117fs*>31 Chr.16: 8855260-8855260 on Build GRCh38
6443837 c.355G>A p.E119K Chr.16: 8855253-8855253 on Build GRCh38
1380604 c.378C>T p.V126V Chr.16: 8855230-8855230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776376647 Chr.16: 8855152-8855152 on Build GRCh38
rs760441738 Chr.16: 8855244-8855244 on Build GRCh38
rs917908391 Chr.16: 8855156-8855156 on Build GRCh38
rs539251894 Chr.16: 8855336-8855337 on Build GRCh38
rs199538832 Chr.16: 8855275-8855275 on Build GRCh38
rs765596310 Chr.16: 8855317-8855317 on Build GRCh38
rs760222054 Chr.16: 8855295-8855295 on Build GRCh38
rs956894677 Chr.16: 8855265-8855265 on Build GRCh38
rs755290520 Chr.16: 8855269-8855269 on Build GRCh38
rs763843068 Chr.16: 8855296-8855296 on Build GRCh38
rs982466247 Chr.16: 8855192-8855192 on Build GRCh38
rs751713938 Chr.16: 8855332-8855332 on Build GRCh38
rs780115934 Chr.16: 8855260-8855260 on Build GRCh38
rs758418358 Chr.16: 8855254-8855254 on Build GRCh38
rs149072869 Chr.16: 8855238-8855238 on Build GRCh38
rs757839359 Chr.16: 8855209-8855209 on Build GRCh38
rs765801909 Chr.16: 8855323-8855324 on Build GRCh38
rs374009548 Chr.16: 8855230-8855230 on Build GRCh38
rs962797665 Chr.16: 8855206-8855206 on Build GRCh38
rs755124372 Chr.16: 8855333-8855333 on Build GRCh38
rs766437174 Chr.16: 8855331-8855331 on Build GRCh38
rs960866702 Chr.16: 8855176-8855176 on Build GRCh38
rs201523233 Chr.16: 8855292-8855292 on Build GRCh38
rs1052964759 Chr.16: 8855245-8855245 on Build GRCh38
rs775196893 Chr.16: 8855293-8855293 on Build GRCh38
rs754812351 Chr.16: 8855342-8855343 on Build GRCh38
rs762186523 Chr.16: 8855313-8855313 on Build GRCh38
rs762955858 Chr.16: 8855170-8855170 on Build GRCh38
rs746072869 Chr.16: 8855142-8855142 on Build GRCh38
rs769814703 Chr.16: 8855211-8855211 on Build GRCh38
rs147662412 Chr.16: 8855204-8855204 on Build GRCh38
rs759243684 Chr.16: 8855186-8855186 on Build GRCh38
rs747231593 Chr.16: 8855262-8855262 on Build GRCh38
rs148643770 Chr.16: 8855247-8855247 on Build GRCh38
rs561939972 Chr.16: 8855303-8855303 on Build GRCh38
rs373801670 Chr.16: 8855270-8855270 on Build GRCh38
rs761553234 Chr.16: 8855159-8855159 on Build GRCh38
rs765806303 Chr.16: 8855180-8855180 on Build GRCh38
rs750865919 Chr.16: 8855323-8855323 on Build GRCh38
rs767320978 Chr.16: 8855191-8855191 on Build GRCh38
rs749148207 Chr.16: 8855272-8855272 on Build GRCh38
rs769411731 Chr.16: 8855229-8855229 on Build GRCh38
rs772166654 Chr.16: 8855222-8855222 on Build GRCh38
rs538591141 Chr.16: 8855257-8855257 on Build GRCh38
rs150215982 Chr.16: 8855232-8855232 on Build GRCh38
rs748342615 Chr.16: 8855342-8855342 on Build GRCh38
rs142480707 Chr.16: 8855226-8855226 on Build GRCh38
rs772063723 Chr.16: 8855148-8855148 on Build GRCh38
rs369895461 Chr.16: 8855200-8855200 on Build GRCh38
rs138038827 Chr.16: 8855234-8855234 on Build GRCh38
rs774383390 Chr.16: 8855276-8855276 on Build GRCh38
rs781436657 Chr.16: 8855339-8855339 on Build GRCh38
rs745983108 Chr.16: 8855217-8855217 on Build GRCh38
rs138738390 Chr.16: 8855253-8855253 on Build GRCh38
rs770848683 Chr.16: 8855140-8855140 on Build GRCh38
rs201509310 Chr.16: 8855239-8855239 on Build GRCh38
rs752431589 Chr.16: 8855198-8855198 on Build GRCh38
rs960835413 Chr.16: 8855338-8855338 on Build GRCh38
rs770951167 Chr.16: 8855233-8855233 on Build GRCh38
rs772968664 Chr.16: 8855168-8855168 on Build GRCh38
rs769656308 Chr.16: 8855163-8855163 on Build GRCh38
rs765690043 Chr.16: 8855252-8855252 on Build GRCh38
rs2231709 Chr.16: 8855330-8855330 on Build GRCh38
rs200524268 Chr.16: 8855248-8855248 on Build GRCh38
rs992422403 Chr.16: 8855286-8855286 on Build GRCh38
rs745803365 Chr.16: 8855277-8855277 on Build GRCh38
rs758109026 Chr.16: 8855201-8855225 on Build GRCh38
rs751172768 Chr.16: 8855181-8855181 on Build GRCh38
rs778202120 Chr.16: 8855207-8855207 on Build GRCh38

View Full Product Details