Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 22945969-22946240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM126A
Entrez Gene ID
84668
Gene Name
family with sequence similarity 126 member A
Gene Aliases
DRCTNNB1A, HCC, HLD5, HYCC1
UniGene
Hs.85603
Gene Chromosome Location
Chr.7: 22895848-23014195 on Build GRCh38
Transcripts
NM_032581.3 XM_005249894.3 XM_005249895.2 XM_011515589.2 XM_011515590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
421498 c.1179A>G p.E393E Chr.7: 22945976-22945976 on Build GRCh38
5002450 c.1001A>G p.E334G Chr.7: 22946154-22946154 on Build GRCh38
3674902 c.1128T>C p.S376S Chr.7: 22946027-22946027 on Build GRCh38
6574793 c.1036G>A p.E346K Chr.7: 22946119-22946119 on Build GRCh38
600597 c.1086G>T p.S362S Chr.7: 22946069-22946069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752176038 Chr.7: 22946088-22946088 on Build GRCh38
rs764219815 Chr.7: 22946087-22946087 on Build GRCh38
rs374199558 Chr.7: 22946148-22946148 on Build GRCh38
rs761145982 Chr.7: 22946144-22946144 on Build GRCh38
rs373770623 Chr.7: 22946023-22946023 on Build GRCh38
rs138563857 Chr.7: 22946072-22946072 on Build GRCh38
rs146436512 Chr.7: 22946007-22946007 on Build GRCh38
rs771923181 Chr.7: 22946069-22946069 on Build GRCh38
rs552434997 Chr.7: 22946167-22946167 on Build GRCh38
rs746845808 Chr.7: 22946185-22946185 on Build GRCh38
rs184953106 Chr.7: 22946178-22946178 on Build GRCh38
rs748068171 Chr.7: 22946066-22946066 on Build GRCh38
rs1043003486 Chr.7: 22946067-22946067 on Build GRCh38
rs770736493 Chr.7: 22946186-22946186 on Build GRCh38
rs750836723 Chr.7: 22946115-22946115 on Build GRCh38
rs750797954 Chr.7: 22946031-22946031 on Build GRCh38
rs947263769 Chr.7: 22946056-22946056 on Build GRCh38
rs910400018 Chr.7: 22946020-22946020 on Build GRCh38
rs756465336 Chr.7: 22946131-22946131 on Build GRCh38
rs914289843 Chr.7: 22946092-22946092 on Build GRCh38
rs777639929 Chr.7: 22946184-22946184 on Build GRCh38
rs753511775 Chr.7: 22946161-22946161 on Build GRCh38
rs778391159 Chr.7: 22946169-22946169 on Build GRCh38
rs767833263 Chr.7: 22946189-22946194 on Build GRCh38
rs922974640 Chr.7: 22946152-22946152 on Build GRCh38
rs755837969 Chr.7: 22946026-22946026 on Build GRCh38
rs757752437 Chr.7: 22946008-22946008 on Build GRCh38
rs774207763 Chr.7: 22946043-22946043 on Build GRCh38
rs140798686 Chr.7: 22946032-22946032 on Build GRCh38
rs1034243737 Chr.7: 22946040-22946040 on Build GRCh38
rs775632203 Chr.7: 22946203-22946203 on Build GRCh38
rs768075744 Chr.7: 22946111-22946111 on Build GRCh38
rs761918014 Chr.7: 22946169-22946169 on Build GRCh38
rs775750049 Chr.7: 22946080-22946080 on Build GRCh38
rs746165960 Chr.7: 22946025-22946025 on Build GRCh38
rs764038282 Chr.7: 22946004-22946006 on Build GRCh38
rs751699541 Chr.7: 22946048-22946049 on Build GRCh38
rs762991662 Chr.7: 22946083-22946083 on Build GRCh38
rs779704756 Chr.7: 22946028-22946028 on Build GRCh38
rs144706867 Chr.7: 22946070-22946070 on Build GRCh38
rs754508003 Chr.7: 22946165-22946165 on Build GRCh38
rs115568145 Chr.7: 22946091-22946091 on Build GRCh38
rs115772574 Chr.7: 22946174-22946174 on Build GRCh38
rs764549315 Chr.7: 22945998-22945998 on Build GRCh38
rs1002489694 Chr.7: 22946033-22946033 on Build GRCh38
rs977059443 Chr.7: 22946134-22946134 on Build GRCh38

View Full Product Details