Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3761278-3761509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC27
Entrez Gene ID
148870
Gene Name
coiled-coil domain containing 27
Gene Aliases
-
UniGene
Hs.348700
Gene Chromosome Location
Chr.1: 3752398-3771645 on Build GRCh38
Transcripts
NM_152492.2 XM_011540780.1 XM_017000402.1 XM_017000403.1 XM_017000404.1 XM_017000405.1 XM_017000406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3805077 c.725C>T p.S242F Chr.1: 3761294-3761294 on Build GRCh38
6063240 c.791C>T p.A264V Chr.1: 3761360-3761360 on Build GRCh38
3369734 c.787G>A p.E263K Chr.1: 3761356-3761356 on Build GRCh38
7086581 c.741G>A p.Q247Q Chr.1: 3761310-3761310 on Build GRCh38
5380839 c.823G>A p.G275S Chr.1: 3761392-3761392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766910113 Chr.1: 3761378-3761378 on Build GRCh38
rs762361664 Chr.1: 3761464-3761464 on Build GRCh38
rs781767309 Chr.1: 3761345-3761347 on Build GRCh38
rs758006720 Chr.1: 3761410-3761410 on Build GRCh38
rs1181883 Chr.1: 3761369-3761369 on Build GRCh38
rs764686407 Chr.1: 3761399-3761399 on Build GRCh38
rs750788282 Chr.1: 3761486-3761486 on Build GRCh38
rs781329951 Chr.1: 3761426-3761426 on Build GRCh38
rs752296471 Chr.1: 3761305-3761305 on Build GRCh38
rs762056417 Chr.1: 3761356-3761356 on Build GRCh38
rs939893498 Chr.1: 3761431-3761431 on Build GRCh38
rs777275065 Chr.1: 3761413-3761413 on Build GRCh38
rs754319019 Chr.1: 3761382-3761382 on Build GRCh38
rs937946531 Chr.1: 3761454-3761454 on Build GRCh38
rs757145057 Chr.1: 3761476-3761476 on Build GRCh38
rs143408613 Chr.1: 3761419-3761419 on Build GRCh38
rs774748457 Chr.1: 3761471-3761471 on Build GRCh38
rs570903636 Chr.1: 3761441-3761441 on Build GRCh38
rs375551418 Chr.1: 3761472-3761472 on Build GRCh38
rs745756388 Chr.1: 3761430-3761430 on Build GRCh38
rs772398433 Chr.1: 3761442-3761442 on Build GRCh38
rs200698032 Chr.1: 3761334-3761334 on Build GRCh38
rs759069493 Chr.1: 3761388-3761388 on Build GRCh38
rs752347360 Chr.1: 3761408-3761408 on Build GRCh38
rs371215327 Chr.1: 3761457-3761457 on Build GRCh38
rs749421623 Chr.1: 3761337-3761337 on Build GRCh38
rs529112190 Chr.1: 3761358-3761358 on Build GRCh38
rs749661908 Chr.1: 3761427-3761427 on Build GRCh38
rs565682717 Chr.1: 3761370-3761370 on Build GRCh38
rs774740825 Chr.1: 3761339-3761339 on Build GRCh38
rs768891256 Chr.1: 3761435-3761435 on Build GRCh38
rs377329678 Chr.1: 3761456-3761456 on Build GRCh38
rs773304302 Chr.1: 3761446-3761446 on Build GRCh38
rs777553154 Chr.1: 3761325-3761325 on Build GRCh38
rs771234548 Chr.1: 3761448-3761448 on Build GRCh38
rs370631959 Chr.1: 3761326-3761326 on Build GRCh38
rs758155423 Chr.1: 3761324-3761324 on Build GRCh38
rs886932440 Chr.1: 3761368-3761368 on Build GRCh38
rs148373212 Chr.1: 3761436-3761436 on Build GRCh38
rs775121904 Chr.1: 3761460-3761460 on Build GRCh38
rs539571122 Chr.1: 3761447-3761447 on Build GRCh38
rs529586550 Chr.1: 3761484-3761484 on Build GRCh38
rs771415587 Chr.1: 3761437-3761437 on Build GRCh38
rs777153852 Chr.1: 3761453-3761453 on Build GRCh38
rs899518219 Chr.1: 3761376-3761376 on Build GRCh38
rs868071802 Chr.1: 3761433-3761433 on Build GRCh38

View Full Product Details