Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129171391-129171644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNBP
Entrez Gene ID
7555
Gene Name
CCHC-type zinc finger nucleic acid binding protein
Gene Aliases
CNBP1, DM2, PROMM, RNF163, ZCCHC22, ZNF9
UniGene
Hs.518249
Gene Chromosome Location
Chr.3: 129167815-129183967 on Build GRCh38
Transcripts
NM_001127192.1 NM_001127193.1 NM_001127194.1 NM_001127195.1 NM_001127196.1 NM_003418.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1186402 c.170G>T p.C57F Chr.3: 129171493-129171493 on Build GRCh38
4630827 c.117G>A p.S39S Chr.3: 129171641-129171641 on Build GRCh38
7361678 c.212A>C p.E71A Chr.3: 129171451-129171451 on Build GRCh38
1191949 c.148C>G p.L50V Chr.3: 129171515-129171515 on Build GRCh38
5042178 c.154G>A p.D52N Chr.3: 129171509-129171509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368927297 Chr.3: 129171427-129171427 on Build GRCh38
rs915643340 Chr.3: 129171498-129171498 on Build GRCh38
rs751743604 Chr.3: 129171591-129171591 on Build GRCh38
rs148679725 Chr.3: 129171577-129171577 on Build GRCh38
rs933380967 Chr.3: 129171554-129171554 on Build GRCh38
rs923348047 Chr.3: 129171541-129171541 on Build GRCh38
rs777925775 Chr.3: 129171473-129171473 on Build GRCh38
rs375834867 Chr.3: 129171574-129171574 on Build GRCh38
rs758364031 Chr.3: 129171472-129171472 on Build GRCh38
rs911819120 Chr.3: 129171594-129171594 on Build GRCh38
rs754261539 Chr.3: 129171418-129171418 on Build GRCh38
rs988173206 Chr.3: 129171540-129171540 on Build GRCh38
rs147219366 Chr.3: 129171568-129171568 on Build GRCh38
rs367980822 Chr.3: 129171519-129171519 on Build GRCh38
rs4303883 Chr.3: 129171507-129171507 on Build GRCh38
rs796494981 Chr.3: 129171437-129171437 on Build GRCh38
rs757328497 Chr.3: 129171505-129171505 on Build GRCh38
rs531181927 Chr.3: 129171608-129171608 on Build GRCh38
rs370108737 Chr.3: 129171586-129171586 on Build GRCh38
rs112583880 Chr.3: 129171445-129171445 on Build GRCh38
rs767991238 Chr.3: 129171545-129171545 on Build GRCh38
rs771479532 Chr.3: 129171565-129171565 on Build GRCh38
rs769536667 Chr.3: 129171512-129171512 on Build GRCh38
rs781315240 Chr.3: 129171600-129171600 on Build GRCh38
rs367731601 Chr.3: 129171564-129171564 on Build GRCh38
rs757235350 Chr.3: 129171599-129171599 on Build GRCh38
rs755815162 Chr.3: 129171610-129171610 on Build GRCh38
rs185055616 Chr.3: 129171559-129171559 on Build GRCh38
rs112103722 Chr.3: 129171444-129171444 on Build GRCh38
rs765419161 Chr.3: 129171436-129171436 on Build GRCh38
rs199646134 Chr.3: 129171588-129171588 on Build GRCh38
rs761183015 Chr.3: 129171413-129171413 on Build GRCh38
rs534922302 Chr.3: 129171570-129171570 on Build GRCh38
rs762582749 Chr.3: 129171501-129171501 on Build GRCh38
rs11542606 Chr.3: 129171487-129171487 on Build GRCh38
rs766662321 Chr.3: 129171417-129171417 on Build GRCh38
rs779532978 Chr.3: 129171522-129171522 on Build GRCh38
rs752998924 Chr.3: 129171441-129171441 on Build GRCh38
rs752808371 Chr.3: 129171584-129171584 on Build GRCh38

View Full Product Details