Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114581626-114581885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPE2
Entrez Gene ID
120406
Gene Name
neurexophilin and PC-esterase domain family member 2
Gene Aliases
FAM55B
UniGene
Hs.446760
Gene Chromosome Location
Chr.11: 114464276-114813702 on Build GRCh38
Transcripts
XM_017017207.1 XM_017017208.1 XM_017017209.1 XM_017017210.1 NM_001077639.1 NM_017678.2 XM_006718865.3 XM_011542881.2 XM_011542882.2 XM_011542883.2

Gene Symbol
NXPE4
Entrez Gene ID
54827
Gene Name
neurexophilin and PC-esterase domain family member 4
Gene Aliases
C11orf33, FAM55D
UniGene
Hs.179100
Gene Chromosome Location
Chr.11: 114570591-114596215 on Build GRCh38
Transcripts
XM_017017207.1 XM_017017208.1 XM_017017209.1 XM_017017210.1 NM_001077639.1 NM_017678.2 XM_006718865.3 XM_011542881.2 XM_011542882.2 XM_011542883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5031272 c.849G>A p.E283E Chr.11: 114581768-114581768 on Build GRCh38
1351436 c.876T>C p.S292S Chr.11: 114581741-114581741 on Build GRCh38
5031273 c.833C>G p.S278* Chr.11: 114581784-114581784 on Build GRCh38
6731962 c.867T>C p.N289N Chr.11: 114581750-114581750 on Build GRCh38
7260893 c.860A>C p.K287T Chr.11: 114581757-114581757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765501365 Chr.11: 114581826-114581826 on Build GRCh38
rs998340563 Chr.11: 114581811-114581811 on Build GRCh38
rs373358602 Chr.11: 114581820-114581820 on Build GRCh38
rs956799029 Chr.11: 114581654-114581654 on Build GRCh38
rs770441734 Chr.11: 114581781-114581781 on Build GRCh38
rs370325500 Chr.11: 114581802-114581802 on Build GRCh38
rs368899366 Chr.11: 114581664-114581664 on Build GRCh38
rs745728625 Chr.11: 114581840-114581840 on Build GRCh38
rs751352671 Chr.11: 114581711-114581711 on Build GRCh38
rs753784668 Chr.11: 114581688-114581688 on Build GRCh38
rs761065343 Chr.11: 114581809-114581809 on Build GRCh38
rs1034668567 Chr.11: 114581794-114581794 on Build GRCh38
rs556201239 Chr.11: 114581766-114581766 on Build GRCh38
rs772423407 Chr.11: 114581803-114581803 on Build GRCh38
rs1037164597 Chr.11: 114581795-114581795 on Build GRCh38
rs779231112 Chr.11: 114581702-114581702 on Build GRCh38
rs759453866 Chr.11: 114581835-114581835 on Build GRCh38
rs112956057 Chr.11: 114581690-114581690 on Build GRCh38
rs748860698 Chr.11: 114581773-114581773 on Build GRCh38
rs769815002 Chr.11: 114581750-114581750 on Build GRCh38
rs759827583 Chr.11: 114581787-114581787 on Build GRCh38
rs776378635 Chr.11: 114581741-114581741 on Build GRCh38
rs879213368 Chr.11: 114581684-114581684 on Build GRCh38
rs757390564 Chr.11: 114581699-114581699 on Build GRCh38
rs748083365 Chr.11: 114581742-114581742 on Build GRCh38
rs927868674 Chr.11: 114581682-114581682 on Build GRCh38
rs112204071 Chr.11: 114581683-114581683 on Build GRCh38
rs138646343 Chr.11: 114581824-114581824 on Build GRCh38
rs764068948 Chr.11: 114581687-114581687 on Build GRCh38
rs1020121072 Chr.11: 114581672-114581672 on Build GRCh38
rs909502889 Chr.11: 114581763-114581763 on Build GRCh38
rs770905307 Chr.11: 114581736-114581736 on Build GRCh38
rs764703175 Chr.11: 114581818-114581818 on Build GRCh38
rs754837312 Chr.11: 114581725-114581725 on Build GRCh38
rs775892676 Chr.11: 114581804-114581804 on Build GRCh38
rs774003980 Chr.11: 114581785-114581785 on Build GRCh38

View Full Product Details