Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207567468-207567725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB1
Entrez Gene ID
1385
Gene Name
cAMP responsive element binding protein 1
Gene Aliases
CREB, CREB-1
UniGene
Hs.516646
Gene Chromosome Location
Chr.2: 207529892-207605989 on Build GRCh38
Transcripts
NM_001320793.1 NM_004379.4 NM_134442.4 NR_135473.1 XM_011510645.1 XM_011510646.2 XM_011510647.2 XM_011510648.2 XM_011510649.2 XM_011510650.2 XM_011510651.2 XM_017003399.1 XM_017003400.1 XM_017003401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6635282 c.350G>A p.S117N Chr.2: 207567509-207567509 on Build GRCh38
4090626 c.370C>T p.R124* Chr.2: 207567529-207567529 on Build GRCh38
160200 c.325G>A p.E109K Chr.2: 207567484-207567484 on Build GRCh38
1015519 c.336G>T p.Q112H Chr.2: 207567495-207567495 on Build GRCh38
3709362 c.374G>C p.R125T Chr.2: 207567533-207567533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952823703 Chr.2: 207567664-207567664 on Build GRCh38
rs558336000 Chr.2: 207567654-207567654 on Build GRCh38
rs540098128 Chr.2: 207567641-207567641 on Build GRCh38
rs745325596 Chr.2: 207567521-207567521 on Build GRCh38
rs774548119 Chr.2: 207567612-207567612 on Build GRCh38
rs909841301 Chr.2: 207567635-207567635 on Build GRCh38
rs768413791 Chr.2: 207567594-207567594 on Build GRCh38
rs941430761 Chr.2: 207567694-207567694 on Build GRCh38
rs387906617 Chr.2: 207567506-207567506 on Build GRCh38
rs751576701 Chr.2: 207567571-207567571 on Build GRCh38
rs769443126 Chr.2: 207567552-207567552 on Build GRCh38
rs200075241 Chr.2: 207567613-207567613 on Build GRCh38
rs555498571 Chr.2: 207567569-207567569 on Build GRCh38
rs3730276 Chr.2: 207567622-207567622 on Build GRCh38
rs894242461 Chr.2: 207567609-207567609 on Build GRCh38

View Full Product Details