Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68749447-68749692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCAR1
Entrez Gene ID
55749
Gene Name
cell division cycle and apoptosis regulator 1
Gene Aliases
-
UniGene
Hs.49853
Gene Chromosome Location
Chr.10: 68721144-68792377 on Build GRCh38
Transcripts
NM_001282959.1 NM_001282960.1 NM_018237.3 NR_104262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
388935 c.1049G>T p.R350L Chr.10: 68749616-68749616 on Build GRCh38
1474721 c.961_962delGA p.E325fs*3 Chr.10: 68749528-68749529 on Build GRCh38
7325766 c.1042C>T p.R348* Chr.10: 68749609-68749609 on Build GRCh38
6299856 c.1082G>T p.R361L Chr.10: 68749649-68749649 on Build GRCh38
685554 c.1093C>T p.Q365* Chr.10: 68749660-68749660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751888685 Chr.10: 68749629-68749629 on Build GRCh38
rs974253861 Chr.10: 68749484-68749484 on Build GRCh38
rs755201637 Chr.10: 68749526-68749526 on Build GRCh38
rs778834570 Chr.10: 68749554-68749554 on Build GRCh38
rs745859960 Chr.10: 68749564-68749564 on Build GRCh38
rs767729481 Chr.10: 68749637-68749637 on Build GRCh38
rs772016049 Chr.10: 68749577-68749577 on Build GRCh38
rs776357944 Chr.10: 68749600-68749600 on Build GRCh38
rs768457972 Chr.10: 68749588-68749588 on Build GRCh38
rs752858433 Chr.10: 68749648-68749648 on Build GRCh38
rs775200198 Chr.10: 68749585-68749585 on Build GRCh38
rs147413396 Chr.10: 68749528-68749529 on Build GRCh38
rs748256307 Chr.10: 68749547-68749547 on Build GRCh38
rs930116694 Chr.10: 68749520-68749520 on Build GRCh38
rs143403057 Chr.10: 68749624-68749624 on Build GRCh38
rs763406750 Chr.10: 68749617-68749617 on Build GRCh38
rs781341103 Chr.10: 68749527-68749527 on Build GRCh38
rs766304257 Chr.10: 68749525-68749525 on Build GRCh38
rs928670048 Chr.10: 68749579-68749579 on Build GRCh38
rs1031482580 Chr.10: 68749645-68749645 on Build GRCh38
rs764902051 Chr.10: 68749610-68749610 on Build GRCh38
rs1000966819 Chr.10: 68749601-68749601 on Build GRCh38
rs756315209 Chr.10: 68749656-68749656 on Build GRCh38
rs756119233 Chr.10: 68749553-68749553 on Build GRCh38
rs750842330 Chr.10: 68749481-68749481 on Build GRCh38
rs746771621 Chr.10: 68749586-68749586 on Build GRCh38
rs980226679 Chr.10: 68749527-68749528 on Build GRCh38
rs759852577 Chr.10: 68749630-68749630 on Build GRCh38
rs184581048 Chr.10: 68749518-68749518 on Build GRCh38
rs772770201 Chr.10: 68749616-68749616 on Build GRCh38
rs758934978 Chr.10: 68749485-68749485 on Build GRCh38
rs942714982 Chr.10: 68749563-68749563 on Build GRCh38
rs1039727484 Chr.10: 68749580-68749580 on Build GRCh38
rs752836010 Chr.10: 68749609-68749609 on Build GRCh38
rs765846105 Chr.10: 68749480-68749480 on Build GRCh38

View Full Product Details