Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6232557-6232814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD9
Entrez Gene ID
928
Gene Name
CD9 molecule
Gene Aliases
BTCC-1, DRAP-27, MIC3, MRP-1, TSPAN-29, TSPAN29
UniGene
Hs.114286
Gene Chromosome Location
Chr.12: 6199844-6238271 on Build GRCh38
Transcripts
NM_001769.3 XM_005253814.3 XM_011521040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
942389 c.199G>A p.G67S Chr.12: 6232655-6232655 on Build GRCh38
4617410 c.192C>T p.I64I Chr.12: 6232648-6232648 on Build GRCh38
6623942 c.273G>T p.L91L Chr.12: 6232729-6232729 on Build GRCh38
4430157 c.205C>T p.L69F Chr.12: 6232661-6232661 on Build GRCh38
6838179 c.196G>C p.A66P Chr.12: 6232652-6232652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370989547 Chr.12: 6232632-6232632 on Build GRCh38
rs974267451 Chr.12: 6232658-6232658 on Build GRCh38
rs889933851 Chr.12: 6232760-6232760 on Build GRCh38
rs375979181 Chr.12: 6232776-6232776 on Build GRCh38
rs752621332 Chr.12: 6232760-6232760 on Build GRCh38
rs766579972 Chr.12: 6232743-6232743 on Build GRCh38
rs868080365 Chr.12: 6232686-6232686 on Build GRCh38
rs748035981 Chr.12: 6232736-6232746 on Build GRCh38
rs769977894 Chr.12: 6232693-6232693 on Build GRCh38
rs951497768 Chr.12: 6232740-6232740 on Build GRCh38
rs373755597 Chr.12: 6232605-6232605 on Build GRCh38
rs761926516 Chr.12: 6232592-6232592 on Build GRCh38
rs748870085 Chr.12: 6232628-6232628 on Build GRCh38
rs781053185 Chr.12: 6232610-6232610 on Build GRCh38
rs755620066 Chr.12: 6232623-6232623 on Build GRCh38
rs368308462 Chr.12: 6232712-6232712 on Build GRCh38
rs763480764 Chr.12: 6232783-6232783 on Build GRCh38
rs368540878 Chr.12: 6232629-6232629 on Build GRCh38
rs759261986 Chr.12: 6232749-6232749 on Build GRCh38
rs114003206 Chr.12: 6232684-6232684 on Build GRCh38
rs375598380 Chr.12: 6232633-6232633 on Build GRCh38
rs3181292 Chr.12: 6232602-6232602 on Build GRCh38
rs138825133 Chr.12: 6232681-6232681 on Build GRCh38
rs1000885174 Chr.12: 6232787-6232787 on Build GRCh38
rs766063609 Chr.12: 6232603-6232603 on Build GRCh38
rs113128549 Chr.12: 6232730-6232730 on Build GRCh38
rs777212002 Chr.12: 6232667-6232667 on Build GRCh38
rs767514697 Chr.12: 6232751-6232751 on Build GRCh38
rs750602405 Chr.12: 6232600-6232600 on Build GRCh38
rs753350410 Chr.12: 6232788-6232788 on Build GRCh38
rs115669578 Chr.12: 6232657-6232657 on Build GRCh38
rs763052687 Chr.12: 6232726-6232726 on Build GRCh38
rs747327386 Chr.12: 6232638-6232638 on Build GRCh38
rs201540148 Chr.12: 6232604-6232604 on Build GRCh38
rs944077619 Chr.12: 6232777-6232777 on Build GRCh38
rs909901859 Chr.12: 6232661-6232661 on Build GRCh38
rs374597801 Chr.12: 6232648-6232648 on Build GRCh38
rs150861096 Chr.12: 6232744-6232744 on Build GRCh38
rs765560710 Chr.12: 6232598-6232598 on Build GRCh38
rs866134961 Chr.12: 6232643-6232643 on Build GRCh38
rs377076812 Chr.12: 6232627-6232627 on Build GRCh38
rs752212494 Chr.12: 6232611-6232611 on Build GRCh38
rs773527961 Chr.12: 6232694-6232694 on Build GRCh38

View Full Product Details