Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50953906-50954149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIGD1C
Entrez Gene ID
613227
Gene Name
HIG1 hypoxia inducible domain family member 1C
Gene Aliases
GM921
UniGene
Hs.702849
Gene Chromosome Location
Chr.12: 50936223-50972592 on Build GRCh38
Transcripts
NM_001109619.1 XM_011538649.2 XM_017019783.1 XM_017019784.1 XM_017019785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3792689 c.75C>A p.D25E Chr.12: 50954073-50954073 on Build GRCh38
694311 c.80C>T p.P27L Chr.12: 50954078-50954078 on Build GRCh38
3461836 c.43C>T p.Q15* Chr.12: 50954041-50954041 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1004187468 Chr.12: 50953981-50953981 on Build GRCh38
rs748484636 Chr.12: 50954101-50954101 on Build GRCh38
rs753519310 Chr.12: 50953953-50953953 on Build GRCh38
rs762146905 Chr.12: 50954000-50954000 on Build GRCh38
rs752534834 Chr.12: 50954040-50954040 on Build GRCh38
rs772326502 Chr.12: 50954111-50954111 on Build GRCh38
rs779344500 Chr.12: 50954092-50954092 on Build GRCh38
rs760109968 Chr.12: 50954018-50954018 on Build GRCh38
rs533552893 Chr.12: 50954055-50954055 on Build GRCh38
rs202101626 Chr.12: 50954053-50954053 on Build GRCh38
rs772750440 Chr.12: 50954009-50954009 on Build GRCh38
rs776947824 Chr.12: 50953993-50953993 on Build GRCh38
rs138664769 Chr.12: 50954050-50954050 on Build GRCh38
rs779502507 Chr.12: 50953944-50953944 on Build GRCh38
rs367776986 Chr.12: 50953968-50953968 on Build GRCh38
rs774742206 Chr.12: 50953988-50953988 on Build GRCh38
rs371611101 Chr.12: 50953992-50953992 on Build GRCh38
rs765903222 Chr.12: 50954019-50954019 on Build GRCh38
rs755532661 Chr.12: 50954089-50954089 on Build GRCh38
rs61731820 Chr.12: 50954003-50954003 on Build GRCh38
rs765199188 Chr.12: 50954032-50954032 on Build GRCh38
rs200801540 Chr.12: 50953959-50953959 on Build GRCh38
rs545500673 Chr.12: 50954028-50954028 on Build GRCh38
rs769097305 Chr.12: 50953976-50953976 on Build GRCh38
rs796099674 Chr.12: 50954004-50954004 on Build GRCh38
rs1023636414 Chr.12: 50954069-50954069 on Build GRCh38
rs749415492 Chr.12: 50954085-50954085 on Build GRCh38
rs772878465 Chr.12: 50954113-50954113 on Build GRCh38
rs780307602 Chr.12: 50954077-50954077 on Build GRCh38
rs749847650 Chr.12: 50953970-50953970 on Build GRCh38
rs531251342 Chr.12: 50954051-50954051 on Build GRCh38

View Full Product Details