Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129443919-129444174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRIP2
Entrez Gene ID
57464
Gene Name
striatin interacting protein 2
Gene Aliases
FAM40B, FAR11B
UniGene
Hs.489988
Gene Chromosome Location
Chr.7: 129434433-129488399 on Build GRCh38
Transcripts
NM_001134336.1 NM_020704.2 XM_006716069.3 XM_011516435.1 XM_017012470.1 XM_017012472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6040053 c.208A>G p.S70G Chr.7: 129444032-129444032 on Build GRCh38
744580 c.271C>G p.Q91E Chr.7: 129444095-129444095 on Build GRCh38
6040052 c.208A>G p.S70G Chr.7: 129444032-129444032 on Build GRCh38
4479710 c.234C>T p.T78T Chr.7: 129444058-129444058 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1006338308 Chr.7: 129444131-129444131 on Build GRCh38
rs745598953 Chr.7: 129444013-129444013 on Build GRCh38
rs371441882 Chr.7: 129443990-129443990 on Build GRCh38
rs752418600 Chr.7: 129444048-129444048 on Build GRCh38
rs1010243464 Chr.7: 129443958-129443958 on Build GRCh38
rs774458575 Chr.7: 129444049-129444049 on Build GRCh38
rs781187413 Chr.7: 129444004-129444004 on Build GRCh38
rs768667633 Chr.7: 129444038-129444038 on Build GRCh38
rs902592269 Chr.7: 129443993-129443993 on Build GRCh38
rs760966240 Chr.7: 129444113-129444113 on Build GRCh38
rs201941888 Chr.7: 129444058-129444058 on Build GRCh38
rs532618889 Chr.7: 129443964-129443964 on Build GRCh38
rs770894003 Chr.7: 129443992-129443992 on Build GRCh38
rs957566957 Chr.7: 129443949-129443949 on Build GRCh38
rs748051818 Chr.7: 129444077-129444079 on Build GRCh38
rs376422661 Chr.7: 129443969-129443969 on Build GRCh38
rs752246350 Chr.7: 129444127-129444127 on Build GRCh38
rs768000058 Chr.7: 129444064-129444064 on Build GRCh38
rs763114833 Chr.7: 129444033-129444033 on Build GRCh38
rs565416239 Chr.7: 129443943-129443943 on Build GRCh38
rs571902301 Chr.7: 129443959-129443959 on Build GRCh38
rs775814308 Chr.7: 129444030-129444030 on Build GRCh38
rs562843630 Chr.7: 129444126-129444126 on Build GRCh38
rs996222052 Chr.7: 129444097-129444097 on Build GRCh38
rs1040483780 Chr.7: 129444051-129444051 on Build GRCh38
rs946521981 Chr.7: 129443985-129443985 on Build GRCh38
rs1029642318 Chr.7: 129444105-129444105 on Build GRCh38
rs762568563 Chr.7: 129444031-129444031 on Build GRCh38
rs536493810 Chr.7: 129443966-129443966 on Build GRCh38
rs553486885 Chr.7: 129443942-129443942 on Build GRCh38
rs770142709 Chr.7: 129444029-129444029 on Build GRCh38
rs188667178 Chr.7: 129444093-129444093 on Build GRCh38

View Full Product Details