Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9264167-9264392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H6PD
Entrez Gene ID
9563
Gene Name
hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase
Gene Aliases
CORTRD1, G6PDH, GDH
UniGene
Hs.463511
Gene Chromosome Location
Chr.1: 9234767-9271337 on Build GRCh38
Transcripts
NM_001282587.1 NM_004285.3 XM_005263540.4 XM_006711052.3 XM_017002865.1 XM_017002866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7429234 c.1775_1781delGCTCGAG p.G592fs*125 Chr.1: 9264268-9264274 on Build GRCh38
6677400 c.1769C>T p.S590L Chr.1: 9264262-9264262 on Build GRCh38
3731972 c.1865G>A p.R622H Chr.1: 9264358-9264358 on Build GRCh38
7429233 c.1774_1780delGGCTCGA p.G592fs*125 Chr.1: 9264267-9264273 on Build GRCh38
4474069 c.1892C>T p.S631F Chr.1: 9264385-9264385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759007268 Chr.1: 9264320-9264320 on Build GRCh38
rs781605545 Chr.1: 9264234-9264234 on Build GRCh38
rs772399015 Chr.1: 9264264-9264264 on Build GRCh38
rs112516051 Chr.1: 9264233-9264233 on Build GRCh38
rs200978401 Chr.1: 9264279-9264279 on Build GRCh38
rs17368556 Chr.1: 9264313-9264313 on Build GRCh38
rs780198446 Chr.1: 9264286-9264286 on Build GRCh38
rs139680845 Chr.1: 9264267-9264267 on Build GRCh38
rs774515572 Chr.1: 9264209-9264209 on Build GRCh38
rs606231222 Chr.1: 9264353-9264354 on Build GRCh38
rs146460271 Chr.1: 9264308-9264308 on Build GRCh38
rs1051135788 Chr.1: 9264349-9264349 on Build GRCh38
rs776340923 Chr.1: 9264315-9264315 on Build GRCh38
rs760218420 Chr.1: 9264222-9264222 on Build GRCh38
rs113566793 Chr.1: 9264350-9264350 on Build GRCh38
rs756336095 Chr.1: 9264285-9264285 on Build GRCh38
rs781339773 Chr.1: 9264305-9264305 on Build GRCh38
rs780755970 Chr.1: 9264367-9264367 on Build GRCh38
rs779224543 Chr.1: 9264354-9264354 on Build GRCh38
rs371828494 Chr.1: 9264363-9264363 on Build GRCh38
rs200208026 Chr.1: 9264215-9264215 on Build GRCh38
rs751449332 Chr.1: 9264268-9264268 on Build GRCh38
rs755426644 Chr.1: 9264324-9264325 on Build GRCh38
rs151130360 Chr.1: 9264326-9264326 on Build GRCh38
rs909563421 Chr.1: 9264198-9264198 on Build GRCh38
rs749706862 Chr.1: 9264206-9264206 on Build GRCh38
rs565031451 Chr.1: 9264207-9264207 on Build GRCh38
rs749803685 Chr.1: 9264261-9264261 on Build GRCh38
rs376136980 Chr.1: 9264304-9264304 on Build GRCh38
rs766579471 Chr.1: 9264353-9264353 on Build GRCh38
rs780502365 Chr.1: 9264236-9264257 on Build GRCh38
rs758165914 Chr.1: 9264258-9264258 on Build GRCh38
rs148493577 Chr.1: 9264235-9264235 on Build GRCh38
rs761689644 Chr.1: 9264342-9264342 on Build GRCh38
rs758314088 Chr.1: 9264275-9264275 on Build GRCh38
rs78281451 Chr.1: 9264227-9264227 on Build GRCh38
rs550005185 Chr.1: 9264200-9264200 on Build GRCh38
rs150373672 Chr.1: 9264335-9264335 on Build GRCh38
rs766100678 Chr.1: 9264262-9264262 on Build GRCh38
rs763929208 Chr.1: 9264278-9264278 on Build GRCh38
rs371242953 Chr.1: 9264307-9264307 on Build GRCh38
rs112274890 Chr.1: 9264272-9264272 on Build GRCh38
rs571825467 Chr.1: 9264357-9264357 on Build GRCh38
rs116493689 Chr.1: 9264218-9264218 on Build GRCh38
rs754833034 Chr.1: 9264228-9264228 on Build GRCh38
rs763982488 Chr.1: 9264334-9264334 on Build GRCh38
rs755084471 Chr.1: 9264302-9264302 on Build GRCh38
rs538048443 Chr.1: 9264271-9264271 on Build GRCh38
rs746011937 Chr.1: 9264196-9264196 on Build GRCh38
rs373847789 Chr.1: 9264356-9264356 on Build GRCh38
rs904627027 Chr.1: 9264322-9264322 on Build GRCh38
rs201127379 Chr.1: 9264223-9264223 on Build GRCh38
rs752491509 Chr.1: 9264232-9264232 on Build GRCh38
rs749388425 Chr.1: 9264290-9264290 on Build GRCh38
rs745471447 Chr.1: 9264370-9264370 on Build GRCh38
rs149851560 Chr.1: 9264263-9264263 on Build GRCh38
rs752649298 Chr.1: 9264273-9264273 on Build GRCh38
rs549607178 Chr.1: 9264257-9264257 on Build GRCh38
rs761359109 Chr.1: 9264231-9264231 on Build GRCh38
rs368005946 Chr.1: 9264358-9264358 on Build GRCh38
rs769215400 Chr.1: 9264373-9264373 on Build GRCh38
rs200495395 Chr.1: 9264241-9264241 on Build GRCh38
rs781550671 Chr.1: 9264194-9264194 on Build GRCh38
rs771294762 Chr.1: 9264364-9264364 on Build GRCh38
rs749816055 Chr.1: 9264262-9264263 on Build GRCh38
rs771217668 Chr.1: 9264186-9264186 on Build GRCh38
rs538653936 Chr.1: 9264281-9264281 on Build GRCh38
rs143602062 Chr.1: 9264362-9264362 on Build GRCh38
rs149313646 Chr.1: 9264216-9264216 on Build GRCh38

View Full Product Details