Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126549943-126550200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf22
Entrez Gene ID
152065
Gene Name
chromosome 3 open reading frame 22
Gene Aliases
-
UniGene
Hs.178210
Gene Chromosome Location
Chr.3: 126524439-126558965 on Build GRCh38
Transcripts
NM_152533.2 NR_130715.1 XM_011512455.2 XM_011512457.2 XM_017005751.1 XM_017005752.1 XM_017005753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616005 c.333_334insC p.R112fs*13 Chr.3: 126549960-126549961 on Build GRCh38
1037782 c.277T>G p.S93A Chr.3: 126550017-126550017 on Build GRCh38
6616006 c.326G>A p.R109H Chr.3: 126549968-126549968 on Build GRCh38
1037781 c.322C>T p.R108C Chr.3: 126549972-126549972 on Build GRCh38
6237969 c.280C>A p.P94T Chr.3: 126550014-126550014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764055133 Chr.3: 126549998-126549998 on Build GRCh38
rs781051967 Chr.3: 126550094-126550095 on Build GRCh38
rs753155117 Chr.3: 126550034-126550034 on Build GRCh38
rs771275774 Chr.3: 126550037-126550037 on Build GRCh38
rs762729878 Chr.3: 126549972-126549972 on Build GRCh38
rs1000316401 Chr.3: 126550122-126550122 on Build GRCh38
rs972629278 Chr.3: 126549976-126549976 on Build GRCh38
rs869463 Chr.3: 126550075-126550075 on Build GRCh38
rs781344616 Chr.3: 126550038-126550038 on Build GRCh38
rs113303966 Chr.3: 126550036-126550036 on Build GRCh38
rs761603029 Chr.3: 126550017-126550017 on Build GRCh38
rs781543989 Chr.3: 126550111-126550111 on Build GRCh38
rs766184972 Chr.3: 126550106-126550106 on Build GRCh38
rs767062392 Chr.3: 126550018-126550018 on Build GRCh38
rs376064235 Chr.3: 126550101-126550101 on Build GRCh38
rs779851364 Chr.3: 126550033-126550033 on Build GRCh38
rs755891744 Chr.3: 126550032-126550032 on Build GRCh38
rs749127935 Chr.3: 126550074-126550074 on Build GRCh38
rs202039278 Chr.3: 126549971-126549971 on Build GRCh38
rs186693615 Chr.3: 126550112-126550112 on Build GRCh38
rs750623054 Chr.3: 126549964-126549964 on Build GRCh38
rs869464 Chr.3: 126550110-126550110 on Build GRCh38
rs958961287 Chr.3: 126550164-126550164 on Build GRCh38
rs775655775 Chr.3: 126550071-126550071 on Build GRCh38
rs560263941 Chr.3: 126550144-126550144 on Build GRCh38
rs554130404 Chr.3: 126549992-126549992 on Build GRCh38
rs751488553 Chr.3: 126550001-126550001 on Build GRCh38
rs1007221821 Chr.3: 126550063-126550063 on Build GRCh38
rs773972987 Chr.3: 126550081-126550081 on Build GRCh38
rs761827585 Chr.3: 126550088-126550088 on Build GRCh38
rs373556472 Chr.3: 126549967-126549967 on Build GRCh38
rs368609187 Chr.3: 126550107-126550107 on Build GRCh38
rs139410214 Chr.3: 126550046-126550046 on Build GRCh38
rs749849116 Chr.3: 126550020-126550020 on Build GRCh38
rs764524310 Chr.3: 126550109-126550109 on Build GRCh38
rs199990010 Chr.3: 126549968-126549968 on Build GRCh38
rs991471783 Chr.3: 126550169-126550169 on Build GRCh38
rs767346848 Chr.3: 126550093-126550093 on Build GRCh38
rs760233761 Chr.3: 126550102-126550102 on Build GRCh38
rs142130143 Chr.3: 126550035-126550035 on Build GRCh38
rs370438267 Chr.3: 126550043-126550043 on Build GRCh38
rs745932961 Chr.3: 126550039-126550039 on Build GRCh38

View Full Product Details