Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123231235-123231486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf76
Entrez Gene ID
84933
Gene Name
chromosome 8 open reading frame 76
Gene Aliases
-
UniGene
Hs.707401
Gene Chromosome Location
Chr.8: 123219956-123241398 on Build GRCh38
Transcripts
NM_032847.2 NM_001204180.1

Gene Symbol
ZHX1-C8orf76
Entrez Gene ID
100533106
Gene Name
ZHX1-C8orf76 readthrough
Gene Aliases
-
UniGene
Hs.707401
Gene Chromosome Location
Chr.8: 123226189-123274487 on Build GRCh38
Transcripts
NM_032847.2 NM_001204180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1199044 c.803T>G p.F268C Chr.8: 123231312-123231312 on Build GRCh38
1454631 c.761A>G p.E254G Chr.8: 123231354-123231354 on Build GRCh38
159669 c.671C>G p.S224C Chr.8: 123231444-123231444 on Build GRCh38
69046 c.777_787del11 p.T260fs*35 Chr.8: 123231328-123231338 on Build GRCh38
6112078 c.685G>T p.E229* Chr.8: 123231430-123231430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148173181 Chr.8: 123231323-123231323 on Build GRCh38
rs141996688 Chr.8: 123231342-123231342 on Build GRCh38
rs756463901 Chr.8: 123231378-123231378 on Build GRCh38
rs756347830 Chr.8: 123231433-123231433 on Build GRCh38
rs778473330 Chr.8: 123231277-123231277 on Build GRCh38
rs747617746 Chr.8: 123231282-123231282 on Build GRCh38
rs376195153 Chr.8: 123231381-123231381 on Build GRCh38
rs369375046 Chr.8: 123231363-123231363 on Build GRCh38
rs201158151 Chr.8: 123231350-123231350 on Build GRCh38
rs369018257 Chr.8: 123231306-123231306 on Build GRCh38
rs757911549 Chr.8: 123231389-123231389 on Build GRCh38
rs375819680 Chr.8: 123231301-123231301 on Build GRCh38
rs774169274 Chr.8: 123231337-123231340 on Build GRCh38
rs763158198 Chr.8: 123231453-123231453 on Build GRCh38
rs770840453 Chr.8: 123231307-123231307 on Build GRCh38
rs751709027 Chr.8: 123231341-123231341 on Build GRCh38
rs373491057 Chr.8: 123231312-123231312 on Build GRCh38
rs902395846 Chr.8: 123231336-123231336 on Build GRCh38
rs200532140 Chr.8: 123231459-123231459 on Build GRCh38
rs764163963 Chr.8: 123231331-123231331 on Build GRCh38
rs963511561 Chr.8: 123231457-123231458 on Build GRCh38
rs370984961 Chr.8: 123231319-123231319 on Build GRCh38
rs138956285 Chr.8: 123231420-123231420 on Build GRCh38
rs752407109 Chr.8: 123231382-123231382 on Build GRCh38
rs964203137 Chr.8: 123231345-123231345 on Build GRCh38
rs142084336 Chr.8: 123231425-123231425 on Build GRCh38
rs777416927 Chr.8: 123231418-123231418 on Build GRCh38
rs150676502 Chr.8: 123231349-123231349 on Build GRCh38
rs367616509 Chr.8: 123231446-123231446 on Build GRCh38
rs776472476 Chr.8: 123231308-123231308 on Build GRCh38
rs775599408 Chr.8: 123231322-123231322 on Build GRCh38
rs760420077 Chr.8: 123231390-123231390 on Build GRCh38
rs745629177 Chr.8: 123231440-123231440 on Build GRCh38
rs113006115 Chr.8: 123231338-123231338 on Build GRCh38
rs951076805 Chr.8: 123231444-123231444 on Build GRCh38
rs560975368 Chr.8: 123231287-123231287 on Build GRCh38
rs754639610 Chr.8: 123231274-123231274 on Build GRCh38
rs199703332 Chr.8: 123231443-123231443 on Build GRCh38
rs1000288390 Chr.8: 123231325-123231325 on Build GRCh38
rs563937038 Chr.8: 123231426-123231426 on Build GRCh38

View Full Product Details