Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52112330-52112555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L10
Entrez Gene ID
10017
Gene Name
BCL2 like 10
Gene Aliases
BCL-B, Boo, Diva, bcl2-L-10
UniGene
Hs.283672
Gene Chromosome Location
Chr.15: 52109263-52112775 on Build GRCh38
Transcripts
NM_001306168.1 NM_020396.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5748933 c.317C>T p.T106M Chr.15: 52112410-52112410 on Build GRCh38
3401817 c.240G>A p.L80L Chr.15: 52112487-52112487 on Build GRCh38
5553950 c.368T>C p.F123S Chr.15: 52112359-52112359 on Build GRCh38
5697814 c.257T>C p.L86P Chr.15: 52112470-52112470 on Build GRCh38
6391418 c.397G>A p.D133N Chr.15: 52112330-52112330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938540861 Chr.15: 52112466-52112466 on Build GRCh38
rs777133899 Chr.15: 52112498-52112498 on Build GRCh38
rs781619001 Chr.15: 52112418-52112418 on Build GRCh38
rs757355335 Chr.15: 52112390-52112390 on Build GRCh38
rs775611996 Chr.15: 52112522-52112522 on Build GRCh38
rs763077602 Chr.15: 52112472-52112472 on Build GRCh38
rs764336092 Chr.15: 52112384-52112384 on Build GRCh38
rs769447348 Chr.15: 52112350-52112350 on Build GRCh38
rs760671102 Chr.15: 52112525-52112525 on Build GRCh38
rs745673827 Chr.15: 52112510-52112510 on Build GRCh38
rs750711054 Chr.15: 52112469-52112469 on Build GRCh38
rs770818508 Chr.15: 52112439-52112439 on Build GRCh38
rs369936579 Chr.15: 52112465-52112465 on Build GRCh38
rs750575660 Chr.15: 52112400-52112400 on Build GRCh38
rs375623310 Chr.15: 52112507-52112507 on Build GRCh38
rs755670714 Chr.15: 52112491-52112491 on Build GRCh38
rs1042773975 Chr.15: 52112512-52112512 on Build GRCh38
rs769251486 Chr.15: 52112427-52112427 on Build GRCh38
rs202035011 Chr.15: 52112453-52112453 on Build GRCh38
rs2231293 Chr.15: 52112426-52112426 on Build GRCh38
rs780265335 Chr.15: 52112406-52112406 on Build GRCh38
rs370306817 Chr.15: 52112435-52112435 on Build GRCh38
rs781686375 Chr.15: 52112482-52112482 on Build GRCh38
rs767576942 Chr.15: 52112454-52112454 on Build GRCh38
rs137890866 Chr.15: 52112509-52112509 on Build GRCh38
rs1024570641 Chr.15: 52112467-52112467 on Build GRCh38
rs772891269 Chr.15: 52112433-52112433 on Build GRCh38
rs774013602 Chr.15: 52112440-52112440 on Build GRCh38
rs766553014 Chr.15: 52112474-52112474 on Build GRCh38
rs753974167 Chr.15: 52112389-52112389 on Build GRCh38
rs372686033 Chr.15: 52112489-52112489 on Build GRCh38
rs758841562 Chr.15: 52112402-52112402 on Build GRCh38
rs765581405 Chr.15: 52112395-52112395 on Build GRCh38
rs751880588 Chr.15: 52112475-52112475 on Build GRCh38
rs747588045 Chr.15: 52112348-52112348 on Build GRCh38
rs771955647 Chr.15: 52112514-52112514 on Build GRCh38
rs376389256 Chr.15: 52112365-52112365 on Build GRCh38
rs775522397 Chr.15: 52112457-52112457 on Build GRCh38
rs774394999 Chr.15: 52112362-52112362 on Build GRCh38
rs762018336 Chr.15: 52112460-52112460 on Build GRCh38
rs371652431 Chr.15: 52112351-52112351 on Build GRCh38
rs887204641 Chr.15: 52112405-52112405 on Build GRCh38
rs766057327 Chr.15: 52112361-52112361 on Build GRCh38
rs747614100 Chr.15: 52112409-52112409 on Build GRCh38
rs755283260 Chr.15: 52112481-52112481 on Build GRCh38
rs1017725916 Chr.15: 52112391-52112391 on Build GRCh38

View Full Product Details