Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 31146110-31146339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMD
Entrez Gene ID
1756
Gene Name
dystrophin
Gene Aliases
BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85
UniGene
Hs.495912
Gene Chromosome Location
Chr.X: 31119219-33339609 on Build GRCh38
Transcripts
NM_000109.3 NM_004006.2 NM_004009.3 NM_004010.3 NM_004011.3 NM_004012.3 NM_004013.2 NM_004014.2 NM_004015.2 NM_004016.2 NM_004017.2 NM_004018.2 NM_004020.3 NM_004021.2 NM_004022.2 NM_004023.2 XM_006724468.2 XM_006724469.3 XM_006724470.3 XM_006724473.2 XM_006724474.3 XM_006724475.2 XM_011545467.1 XM_017029328.1 XM_017029331.1

SNP Detail

SNP ID Chromosome Location
rs992433271 Chr.X: 31146229-31146229 on Build GRCh38
rs763355870 Chr.X: 31146232-31146232 on Build GRCh38
rs200166885 Chr.X: 31146302-31146302 on Build GRCh38
rs376336151 Chr.X: 31146268-31146268 on Build GRCh38
rs398123848 Chr.X: 31146286-31146286 on Build GRCh38
rs967917321 Chr.X: 31146147-31146147 on Build GRCh38
rs545317905 Chr.X: 31146231-31146231 on Build GRCh38
rs763911015 Chr.X: 31146242-31146243 on Build GRCh38
rs941558150 Chr.X: 31146312-31146312 on Build GRCh38
rs769537502 Chr.X: 31146304-31146304 on Build GRCh38
rs747939733 Chr.X: 31146301-31146301 on Build GRCh38
rs774578079 Chr.X: 31146317-31146317 on Build GRCh38
rs113815302 Chr.X: 31146213-31146213 on Build GRCh38
rs72466536 Chr.X: 31146158-31146158 on Build GRCh38
rs960566725 Chr.X: 31146184-31146184 on Build GRCh38
rs533890798 Chr.X: 31146239-31146239 on Build GRCh38
rs747031916 Chr.X: 31146258-31146258 on Build GRCh38
rs990606875 Chr.X: 31146297-31146297 on Build GRCh38
rs1002860967 Chr.X: 31146212-31146212 on Build GRCh38
rs779397511 Chr.X: 31146251-31146251 on Build GRCh38
rs770447872 Chr.X: 31146183-31146183 on Build GRCh38
rs34651197 Chr.X: 31146289-31146290 on Build GRCh38
rs923736935 Chr.X: 31146165-31146165 on Build GRCh38
rs371742990 Chr.X: 31146224-31146224 on Build GRCh38

View Full Product Details