Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130071375-130071614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELF4
Entrez Gene ID
2000
Gene Name
E74 like ETS transcription factor 4
Gene Aliases
ELFR, MEF
UniGene
Hs.271940
Gene Chromosome Location
Chr.X: 130064920-130110713 on Build GRCh38
Transcripts
NM_001127197.1 NM_001421.3 XM_005262389.3 XM_011531307.2 XM_011531308.2

SNP Detail

SNP ID Chromosome Location
rs948193457 Chr.X: 130071422-130071423 on Build GRCh38
rs202174150 Chr.X: 130071441-130071441 on Build GRCh38
rs1009386799 Chr.X: 130071501-130071501 on Build GRCh38
rs759506295 Chr.X: 130071467-130071467 on Build GRCh38
rs777240185 Chr.X: 130071561-130071561 on Build GRCh38
rs765426369 Chr.X: 130071469-130071469 on Build GRCh38
rs773499524 Chr.X: 130071437-130071437 on Build GRCh38
rs772580240 Chr.X: 130071427-130071427 on Build GRCh38
rs935505498 Chr.X: 130071498-130071498 on Build GRCh38
rs768248298 Chr.X: 130071395-130071395 on Build GRCh38
rs761058400 Chr.X: 130071440-130071440 on Build GRCh38
rs1052519924 Chr.X: 130071577-130071577 on Build GRCh38
rs776961332 Chr.X: 130071456-130071456 on Build GRCh38
rs748858727 Chr.X: 130071593-130071593 on Build GRCh38
rs778108032 Chr.X: 130071490-130071490 on Build GRCh38
rs371941494 Chr.X: 130071470-130071470 on Build GRCh38
rs916548148 Chr.X: 130071400-130071400 on Build GRCh38
rs375750801 Chr.X: 130071398-130071398 on Build GRCh38
rs753175886 Chr.X: 130071432-130071432 on Build GRCh38
rs140747142 Chr.X: 130071417-130071417 on Build GRCh38

View Full Product Details