Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94319648-94319921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUTM2F
Entrez Gene ID
54754
Gene Name
NUT family member 2F
Gene Aliases
FAM22F, NUTMF
UniGene
Hs.648183
Gene Chromosome Location
Chr.9: 94318196-94328644 on Build GRCh38
Transcripts
NM_017561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260793 c.1043G>T p.S348I Chr.9: 94319653-94319653 on Build GRCh38
5530918 c.993C>T p.F331F Chr.9: 94319703-94319703 on Build GRCh38
3659744 c.1033T>C p.L345L Chr.9: 94319663-94319663 on Build GRCh38
3659746 c.997G>A p.E333K Chr.9: 94319699-94319699 on Build GRCh38
5530917 c.1395C>T p.F465F Chr.9: 94319703-94319703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs59383317 Chr.9: 94319721-94319721 on Build GRCh38
rs567044479 Chr.9: 94319685-94319685 on Build GRCh38
rs2257698 Chr.9: 94319813-94319813 on Build GRCh38
rs747488846 Chr.9: 94319720-94319720 on Build GRCh38
rs1000016172 Chr.9: 94319794-94319794 on Build GRCh38
rs756894904 Chr.9: 94319705-94319705 on Build GRCh38
rs776880380 Chr.9: 94319731-94319731 on Build GRCh38
rs777525236 Chr.9: 94319700-94319700 on Build GRCh38
rs1035145596 Chr.9: 94319812-94319812 on Build GRCh38
rs778563985 Chr.9: 94319707-94319707 on Build GRCh38
rs776953246 Chr.9: 94319673-94319673 on Build GRCh38
rs780775742 Chr.9: 94319711-94319711 on Build GRCh38
rs771695770 Chr.9: 94319710-94319710 on Build GRCh38
rs1012423609 Chr.9: 94319865-94319865 on Build GRCh38
rs762196937 Chr.9: 94319737-94319737 on Build GRCh38
rs753623167 Chr.9: 94319701-94319701 on Build GRCh38
rs902537495 Chr.9: 94319793-94319793 on Build GRCh38
rs765450763 Chr.9: 94319679-94319679 on Build GRCh38
rs779923884 Chr.9: 94319760-94319760 on Build GRCh38
rs766439249 Chr.9: 94319770-94319770 on Build GRCh38
rs763075599 Chr.9: 94319765-94319765 on Build GRCh38
rs575433785 Chr.9: 94319845-94319845 on Build GRCh38
rs755258856 Chr.9: 94319859-94319859 on Build GRCh38
rs773421608 Chr.9: 94319760-94319760 on Build GRCh38
rs896622569 Chr.9: 94319832-94319832 on Build GRCh38
rs769140351 Chr.9: 94319671-94319671 on Build GRCh38
rs534291881 Chr.9: 94319708-94319708 on Build GRCh38
rs769911719 Chr.9: 94319746-94319746 on Build GRCh38
rs752615108 Chr.9: 94319690-94319690 on Build GRCh38
rs574123675 Chr.9: 94319795-94319795 on Build GRCh38
rs1044105263 Chr.9: 94319729-94319729 on Build GRCh38
rs763019192 Chr.9: 94319680-94319680 on Build GRCh38
rs28735918 Chr.9: 94319893-94319893 on Build GRCh38
rs557250059 Chr.9: 94319815-94319815 on Build GRCh38
rs1023869449 Chr.9: 94319892-94319892 on Build GRCh38
rs756066222 Chr.9: 94319743-94319745 on Build GRCh38

View Full Product Details