Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 30885960-30886213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COCH
Entrez Gene ID
1690
Gene Name
cochlin
Gene Aliases
COCH-5B2, COCH5B2, DFNA9
UniGene
Hs.21016
Gene Chromosome Location
Chr.14: 30874496-30890618 on Build GRCh38
Transcripts
NM_001135058.1 NM_004086.2 XM_017021071.1 XM_017021072.1 XM_017021073.1 NR_038356.1

Gene Symbol
LOC100506071
Entrez Gene ID
100506071
Gene Name
uncharacterized LOC100506071
Gene Aliases
-
UniGene
Hs.729195
Gene Chromosome Location
Chr.14: 30876179-30889808 on Build GRCh38
Transcripts
NM_001135058.1 NM_004086.2 XM_017021071.1 XM_017021072.1 XM_017021073.1 NR_038356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
84248 c.1199C>T p.S400L Chr.14: 30886034-30886034 on Build GRCh38
5848006 c.752C>T p.S251L Chr.14: 30886034-30886034 on Build GRCh38
5791954 c.821A>C p.D274A Chr.14: 30886103-30886103 on Build GRCh38
5828096 c.750C>A p.I250I Chr.14: 30886032-30886032 on Build GRCh38
1369510 c.1197C>A p.I399I Chr.14: 30886032-30886032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139503327 Chr.14: 30886183-30886183 on Build GRCh38
rs371248806 Chr.14: 30886165-30886165 on Build GRCh38
rs1028873214 Chr.14: 30886125-30886125 on Build GRCh38
rs878853226 Chr.14: 30885994-30885994 on Build GRCh38
rs1008723301 Chr.14: 30886167-30886167 on Build GRCh38
rs776467069 Chr.14: 30886151-30886151 on Build GRCh38
rs768435123 Chr.14: 30886150-30886150 on Build GRCh38
rs775079663 Chr.14: 30886032-30886032 on Build GRCh38
rs367884240 Chr.14: 30886081-30886081 on Build GRCh38
rs571941613 Chr.14: 30885995-30885995 on Build GRCh38
rs776519637 Chr.14: 30886036-30886036 on Build GRCh38
rs772383772 Chr.14: 30886082-30886082 on Build GRCh38
rs748019236 Chr.14: 30886153-30886153 on Build GRCh38
rs199608109 Chr.14: 30886056-30886056 on Build GRCh38
rs1010923717 Chr.14: 30886173-30886173 on Build GRCh38
rs746680829 Chr.14: 30886148-30886148 on Build GRCh38
rs757948966 Chr.14: 30886121-30886121 on Build GRCh38
rs61759484 Chr.14: 30886105-30886105 on Build GRCh38
rs995768209 Chr.14: 30886110-30886110 on Build GRCh38
rs766435124 Chr.14: 30886040-30886040 on Build GRCh38
rs745390437 Chr.14: 30886115-30886115 on Build GRCh38
rs886229195 Chr.14: 30886055-30886055 on Build GRCh38
rs756473964 Chr.14: 30886085-30886085 on Build GRCh38
rs759679894 Chr.14: 30886162-30886162 on Build GRCh38
rs892430382 Chr.14: 30886109-30886109 on Build GRCh38
rs751654014 Chr.14: 30886049-30886049 on Build GRCh38
rs140108742 Chr.14: 30885987-30885987 on Build GRCh38
rs545418366 Chr.14: 30885986-30885986 on Build GRCh38
rs769725783 Chr.14: 30886154-30886154 on Build GRCh38
rs756541797 Chr.14: 30885985-30885985 on Build GRCh38
rs1038336750 Chr.14: 30886012-30886012 on Build GRCh38
rs767814589 Chr.14: 30886074-30886074 on Build GRCh38
rs922013526 Chr.14: 30886015-30886016 on Build GRCh38
rs770530713 Chr.14: 30885991-30885991 on Build GRCh38
rs760549722 Chr.14: 30886034-30886034 on Build GRCh38
rs549957619 Chr.14: 30886086-30886086 on Build GRCh38
rs28362778 Chr.14: 30886039-30886039 on Build GRCh38
rs150529026 Chr.14: 30886035-30886035 on Build GRCh38
rs35353967 Chr.14: 30886104-30886104 on Build GRCh38
rs748478920 Chr.14: 30886052-30886052 on Build GRCh38
rs77370101 Chr.14: 30886002-30886002 on Build GRCh38
rs771767136 Chr.14: 30886023-30886023 on Build GRCh38
rs779778621 Chr.14: 30886145-30886145 on Build GRCh38
rs200904881 Chr.14: 30886019-30886019 on Build GRCh38
rs774252846 Chr.14: 30886159-30886159 on Build GRCh38
rs1020107825 Chr.14: 30886177-30886177 on Build GRCh38

View Full Product Details