Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73495169-73495442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT1
Entrez Gene ID
641371
Gene Name
acyl-CoA thioesterase 1
Gene Aliases
ACH2, CTE-1, LACH2
UniGene
Hs.446685
Gene Chromosome Location
Chr.14: 73493021-73543799 on Build GRCh38
Transcripts
XM_017021590.1 XM_017021591.1 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
UniGene
Hs.660774
Gene Chromosome Location
Chr.14: 73478484-73634418 on Build GRCh38
Transcripts
XM_017021590.1 XM_017021591.1 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5903858 c.2646G>T p.K882N Chr.14: 73495367-73495367 on Build GRCh38
3497791 c.2679G>A p.E893E Chr.14: 73495334-73495334 on Build GRCh38
957643 c.2632G>T p.E878* Chr.14: 73495240-73495240 on Build GRCh38
6311141 c.2740C>A p.Q914K Chr.14: 73495273-73495273 on Build GRCh38
3815264 c.2510T>C p.L837S Chr.14: 73495362-73495362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372881252 Chr.14: 73495412-73495412 on Build GRCh38
rs768636996 Chr.14: 73495327-73495327 on Build GRCh38
rs1057087220 Chr.14: 73495245-73495245 on Build GRCh38
rs761097429 Chr.14: 73495405-73495405 on Build GRCh38
rs774446122 Chr.14: 73495329-73495329 on Build GRCh38
rs371721743 Chr.14: 73495309-73495309 on Build GRCh38
rs866508305 Chr.14: 73495331-73495331 on Build GRCh38
rs745707689 Chr.14: 73495316-73495316 on Build GRCh38
rs745834106 Chr.14: 73495241-73495241 on Build GRCh38
rs772327849 Chr.14: 73495349-73495349 on Build GRCh38
rs34456634 Chr.14: 73495211-73495212 on Build GRCh38
rs756931027 Chr.14: 73495310-73495310 on Build GRCh38
rs200663445 Chr.14: 73495224-73495224 on Build GRCh38
rs750824973 Chr.14: 73495279-73495279 on Build GRCh38
rs780961946 Chr.14: 73495314-73495314 on Build GRCh38
rs747663983 Chr.14: 73495200-73495200 on Build GRCh38
rs757914732 Chr.14: 73495203-73495203 on Build GRCh38
rs774449151 Chr.14: 73495265-73495265 on Build GRCh38
rs775482630 Chr.14: 73495258-73495258 on Build GRCh38
rs374999620 Chr.14: 73495332-73495332 on Build GRCh38
rs201577374 Chr.14: 73495232-73495232 on Build GRCh38
rs369339739 Chr.14: 73495391-73495391 on Build GRCh38
rs754368305 Chr.14: 73495297-73495297 on Build GRCh38
rs143634597 Chr.14: 73495300-73495300 on Build GRCh38
rs138404328 Chr.14: 73495326-73495326 on Build GRCh38
rs1043013871 Chr.14: 73495212-73495212 on Build GRCh38
rs748519813 Chr.14: 73495330-73495330 on Build GRCh38
rs777359042 Chr.14: 73495210-73495210 on Build GRCh38
rs770446372 Chr.14: 73495413-73495417 on Build GRCh38
rs766856357 Chr.14: 73495291-73495291 on Build GRCh38
rs769705232 Chr.14: 73495318-73495318 on Build GRCh38
rs763007867 Chr.14: 73495262-73495262 on Build GRCh38
rs562665737 Chr.14: 73495324-73495324 on Build GRCh38
rs1016568967 Chr.14: 73495290-73495290 on Build GRCh38
rs902708968 Chr.14: 73495238-73495238 on Build GRCh38
rs768714254 Chr.14: 73495263-73495263 on Build GRCh38
rs76429074 Chr.14: 73495278-73495278 on Build GRCh38
rs181918013 Chr.14: 73495256-73495256 on Build GRCh38
rs757855044 Chr.14: 73495299-73495299 on Build GRCh38
rs368885398 Chr.14: 73495282-73495282 on Build GRCh38
rs777881574 Chr.14: 73495412-73495413 on Build GRCh38
rs761970252 Chr.14: 73495271-73495271 on Build GRCh38
rs776519128 Chr.14: 73495237-73495237 on Build GRCh38

View Full Product Details