Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41939558-41939803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO4
Entrez Gene ID
26272
Gene Name
F-box protein 4
Gene Aliases
FBX4
UniGene
Hs.165575
Gene Chromosome Location
Chr.5: 41925249-42040230 on Build GRCh38
Transcripts
NM_001297437.1 NM_012176.2 XM_006714470.3 XM_011514026.2 XM_011514027.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3728066 c.1025T>C p.F342S Chr.5: 41939567-41939567 on Build GRCh38
5531118 c.1057C>T p.L353L Chr.5: 41939599-41939599 on Build GRCh38
7313357 c.1074G>T p.L358L Chr.5: 41939616-41939616 on Build GRCh38
6171318 c.1063C>T p.H355Y Chr.5: 41939605-41939605 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1024916608 Chr.5: 41939680-41939680 on Build GRCh38
rs555700263 Chr.5: 41939679-41939679 on Build GRCh38
rs141978667 Chr.5: 41939754-41939757 on Build GRCh38
rs764618722 Chr.5: 41939716-41939716 on Build GRCh38
rs556998910 Chr.5: 41939762-41939762 on Build GRCh38
rs865850784 Chr.5: 41939755-41939755 on Build GRCh38
rs996522194 Chr.5: 41939586-41939586 on Build GRCh38
rs374556186 Chr.5: 41939622-41939622 on Build GRCh38
rs754092619 Chr.5: 41939610-41939610 on Build GRCh38
rs1051210305 Chr.5: 41939715-41939715 on Build GRCh38
rs61649680 Chr.5: 41939757-41939760 on Build GRCh38
rs971575361 Chr.5: 41939727-41939727 on Build GRCh38
rs766310816 Chr.5: 41939609-41939609 on Build GRCh38
rs1010418125 Chr.5: 41939670-41939670 on Build GRCh38
rs3842033 Chr.5: 41939759-41939762 on Build GRCh38
rs748494197 Chr.5: 41939642-41939642 on Build GRCh38
rs755204377 Chr.5: 41939629-41939629 on Build GRCh38
rs980795543 Chr.5: 41939733-41939733 on Build GRCh38
rs756204103 Chr.5: 41939608-41939608 on Build GRCh38
rs574062197 Chr.5: 41939723-41939723 on Build GRCh38
rs759040665 Chr.5: 41939663-41939663 on Build GRCh38
rs1033060601 Chr.5: 41939656-41939656 on Build GRCh38
rs933144899 Chr.5: 41939677-41939677 on Build GRCh38
rs1006902349 Chr.5: 41939769-41939769 on Build GRCh38
rs779470732 Chr.5: 41939631-41939631 on Build GRCh38
rs541470575 Chr.5: 41939734-41939734 on Build GRCh38
rs750530931 Chr.5: 41939605-41939605 on Build GRCh38
rs1000150655 Chr.5: 41939656-41939656 on Build GRCh38

View Full Product Details