Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 59641179-59641452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGGY
Entrez Gene ID
55277
Gene Name
FGGY carbohydrate kinase domain containing
Gene Aliases
-
UniGene
Hs.444301
Gene Chromosome Location
Chr.1: 59296405-59762730 on Build GRCh38
Transcripts
NM_001113411.1 NM_001244714.1 NM_001278224.1 NM_018291.3 NR_103473.1 XM_011541730.1 XM_011541731.1 XM_011541732.1 XM_011541733.1 XM_011541735.1 XM_011541736.2 XM_017001643.1 XM_017001644.1 XM_017001645.1 XM_017001646.1 XM_017001647.1 XM_017001648.1 XM_017001649.1 XM_017001650.1 XM_017001651.1 XM_017001652.1 XM_017001653.1 XM_017001654.1 XM_017001655.1 XM_017001656.1 XM_017001657.1 XM_017001658.1 XM_017001659.1 XM_017001660.1 XM_017001661.1 XM_017001662.1 XM_017001663.1 XM_017001664.1 XM_017001665.1 XM_017001666.1 XM_017001667.1 XM_017001668.1 XM_017001669.1 XM_017001670.1 XM_017001671.1 XM_017001672.1 XM_017001673.1 XM_017001674.1 XM_017001675.1 XM_017001676.1 XM_017001677.1 XM_017001678.1 XM_017001679.1

SNP Detail

SNP ID Chromosome Location
rs757358287 Chr.1: 59641262-59641262 on Build GRCh38
rs868695938 Chr.1: 59641289-59641290 on Build GRCh38
rs12078813 Chr.1: 59641368-59641368 on Build GRCh38
rs573243673 Chr.1: 59641351-59641351 on Build GRCh38
rs757404709 Chr.1: 59641378-59641378 on Build GRCh38
rs770285878 Chr.1: 59641281-59641281 on Build GRCh38
rs780172010 Chr.1: 59641296-59641296 on Build GRCh38
rs772885646 Chr.1: 59641237-59641241 on Build GRCh38
rs936506502 Chr.1: 59641402-59641402 on Build GRCh38
rs763724953 Chr.1: 59641362-59641362 on Build GRCh38
rs199727905 Chr.1: 59641269-59641269 on Build GRCh38
rs201842494 Chr.1: 59641306-59641306 on Build GRCh38
rs1001643449 Chr.1: 59641212-59641212 on Build GRCh38
rs12036354 Chr.1: 59641258-59641258 on Build GRCh38
rs752632048 Chr.1: 59641347-59641347 on Build GRCh38
rs765190372 Chr.1: 59641234-59641234 on Build GRCh38
rs919511619 Chr.1: 59641297-59641297 on Build GRCh38
rs868131815 Chr.1: 59641290-59641290 on Build GRCh38
rs912421872 Chr.1: 59641371-59641371 on Build GRCh38
rs963898571 Chr.1: 59641293-59641293 on Build GRCh38
rs765241687 Chr.1: 59641330-59641330 on Build GRCh38
rs774320810 Chr.1: 59641298-59641302 on Build GRCh38
rs775532127 Chr.1: 59641383-59641383 on Build GRCh38
rs942518476 Chr.1: 59641404-59641404 on Build GRCh38
rs759435231 Chr.1: 59641302-59641308 on Build GRCh38
rs986242879 Chr.1: 59641277-59641277 on Build GRCh38
rs758741087 Chr.1: 59641246-59641246 on Build GRCh38
rs1030375273 Chr.1: 59641261-59641261 on Build GRCh38
rs747128652 Chr.1: 59641305-59641305 on Build GRCh38
rs376384800 Chr.1: 59641229-59641229 on Build GRCh38
rs776276761 Chr.1: 59641319-59641319 on Build GRCh38
rs558163557 Chr.1: 59641326-59641326 on Build GRCh38
rs778075293 Chr.1: 59641251-59641251 on Build GRCh38
rs975171792 Chr.1: 59641408-59641408 on Build GRCh38
rs759587082 Chr.1: 59641327-59641327 on Build GRCh38
rs1020204469 Chr.1: 59641298-59641298 on Build GRCh38
rs781318684 Chr.1: 59641385-59641385 on Build GRCh38
rs747157058 Chr.1: 59641257-59641257 on Build GRCh38
rs1016320095 Chr.1: 59641283-59641283 on Build GRCh38
rs754951177 Chr.1: 59641232-59641232 on Build GRCh38
rs376687905 Chr.1: 59641299-59641299 on Build GRCh38
rs752507361 Chr.1: 59641243-59641243 on Build GRCh38
rs919660462 Chr.1: 59641423-59641423 on Build GRCh38
rs765926959 Chr.1: 59641318-59641318 on Build GRCh38
rs577625694 Chr.1: 59641244-59641244 on Build GRCh38
rs187153546 Chr.1: 59641317-59641317 on Build GRCh38
rs746435219 Chr.1: 59641276-59641276 on Build GRCh38
rs762767166 Chr.1: 59641256-59641257 on Build GRCh38
rs545619944 Chr.1: 59641291-59641298 on Build GRCh38
rs541717260 Chr.1: 59641328-59641328 on Build GRCh38

View Full Product Details