Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68722390-68722645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCAR1
Entrez Gene ID
55749
Gene Name
cell division cycle and apoptosis regulator 1
Gene Aliases
-
UniGene
Hs.49853
Gene Chromosome Location
Chr.10: 68721144-68792377 on Build GRCh38
Transcripts
NM_001282959.1 NM_001282960.1 NM_018237.3 NR_104262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027239 c.14G>A p.G5E Chr.10: 68722518-68722518 on Build GRCh38
5611940 c.29C>T p.P10L Chr.10: 68722533-68722533 on Build GRCh38
4494786 c.43C>G p.Q15E Chr.10: 68722547-68722547 on Build GRCh38
159777 c.43C>T p.Q15* Chr.10: 68722547-68722547 on Build GRCh38
7203086 c.1A>G p.M1V Chr.10: 68722505-68722505 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371263749 Chr.10: 68722474-68722474 on Build GRCh38
rs757832949 Chr.10: 68722615-68722615 on Build GRCh38
rs746170594 Chr.10: 68722485-68722486 on Build GRCh38
rs778144168 Chr.10: 68722485-68722485 on Build GRCh38
rs963067148 Chr.10: 68722439-68722439 on Build GRCh38
rs751350714 Chr.10: 68722459-68722459 on Build GRCh38
rs771341047 Chr.10: 68722500-68722500 on Build GRCh38
rs61256329 Chr.10: 68722542-68722542 on Build GRCh38
rs988193318 Chr.10: 68722422-68722422 on Build GRCh38
rs746217216 Chr.10: 68722502-68722502 on Build GRCh38
rs780534239 Chr.10: 68722609-68722609 on Build GRCh38
rs777397214 Chr.10: 68722610-68722610 on Build GRCh38
rs749742527 Chr.10: 68722494-68722494 on Build GRCh38
rs764177283 Chr.10: 68722554-68722554 on Build GRCh38
rs374203468 Chr.10: 68722592-68722592 on Build GRCh38
rs777512328 Chr.10: 68722430-68722430 on Build GRCh38
rs781200908 Chr.10: 68722579-68722579 on Build GRCh38
rs766252675 Chr.10: 68722442-68722442 on Build GRCh38
rs979824014 Chr.10: 68722538-68722538 on Build GRCh38
rs377689155 Chr.10: 68722596-68722596 on Build GRCh38
rs373345073 Chr.10: 68722534-68722534 on Build GRCh38
rs375599265 Chr.10: 68722617-68722617 on Build GRCh38
rs113619196 Chr.10: 68722580-68722580 on Build GRCh38
rs760675806 Chr.10: 68722553-68722553 on Build GRCh38
rs758143149 Chr.10: 68722433-68722433 on Build GRCh38
rs775814736 Chr.10: 68722543-68722543 on Build GRCh38
rs767187851 Chr.10: 68722594-68722594 on Build GRCh38
rs754664726 Chr.10: 68722461-68722461 on Build GRCh38
rs375321669 Chr.10: 68722470-68722470 on Build GRCh38
rs908382832 Chr.10: 68722607-68722607 on Build GRCh38
rs200677722 Chr.10: 68722612-68722612 on Build GRCh38
rs369461888 Chr.10: 68722583-68722583 on Build GRCh38
rs202095685 Chr.10: 68722589-68722589 on Build GRCh38
rs912420084 Chr.10: 68722602-68722602 on Build GRCh38
rs770147247 Chr.10: 68722494-68722495 on Build GRCh38
rs750309898 Chr.10: 68722421-68722421 on Build GRCh38
rs369109457 Chr.10: 68722501-68722501 on Build GRCh38
rs755590836 Chr.10: 68722605-68722605 on Build GRCh38
rs113255973 Chr.10: 68722481-68722481 on Build GRCh38

View Full Product Details