Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 298642-298915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFITM5
Entrez Gene ID
387733
Gene Name
interferon induced transmembrane protein 5
Gene Aliases
BRIL, DSPA1, Hrmp1, OI5, fragilis4
UniGene
Hs.443469
Gene Chromosome Location
Chr.11: 298200-299526 on Build GRCh38
Transcripts
NM_001025295.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7371799 c.230G>A p.R77H Chr.11: 298670-298670 on Build GRCh38
4480340 c.240C>T p.S80S Chr.11: 298660-298660 on Build GRCh38
1604463 c.239C>G p.S80C Chr.11: 298661-298661 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755986012 Chr.11: 298673-298673 on Build GRCh38
rs578108064 Chr.11: 298871-298871 on Build GRCh38
rs1038638182 Chr.11: 298840-298840 on Build GRCh38
rs760293020 Chr.11: 298715-298715 on Build GRCh38
rs771867792 Chr.11: 298712-298712 on Build GRCh38
rs755360003 Chr.11: 298760-298760 on Build GRCh38
rs773676058 Chr.11: 298815-298815 on Build GRCh38
rs368090655 Chr.11: 298748-298748 on Build GRCh38
rs759346513 Chr.11: 298731-298731 on Build GRCh38
rs753629362 Chr.11: 298671-298671 on Build GRCh38
rs943039460 Chr.11: 298787-298787 on Build GRCh38
rs374350236 Chr.11: 298674-298674 on Build GRCh38
rs141795168 Chr.11: 298678-298678 on Build GRCh38
rs781543593 Chr.11: 298718-298719 on Build GRCh38
rs933103135 Chr.11: 298882-298882 on Build GRCh38
rs754499637 Chr.11: 298763-298763 on Build GRCh38
rs770229423 Chr.11: 298758-298758 on Build GRCh38
rs538337680 Chr.11: 298771-298771 on Build GRCh38
rs770324603 Chr.11: 298793-298793 on Build GRCh38
rs756231523 Chr.11: 298754-298754 on Build GRCh38
rs757682454 Chr.11: 298676-298678 on Build GRCh38
rs779092529 Chr.11: 298761-298761 on Build GRCh38
rs370167475 Chr.11: 298757-298757 on Build GRCh38
rs777439579 Chr.11: 298747-298748 on Build GRCh38
rs766291648 Chr.11: 298719-298719 on Build GRCh38
rs752549056 Chr.11: 298742-298742 on Build GRCh38
rs760636343 Chr.11: 298663-298663 on Build GRCh38
rs778596622 Chr.11: 298878-298878 on Build GRCh38
rs959655009 Chr.11: 298696-298696 on Build GRCh38
rs757403600 Chr.11: 298747-298747 on Build GRCh38
rs148174338 Chr.11: 298709-298709 on Build GRCh38
rs200872938 Chr.11: 298728-298728 on Build GRCh38
rs947419453 Chr.11: 298798-298798 on Build GRCh38
rs147932991 Chr.11: 298670-298670 on Build GRCh38
rs2293745 Chr.11: 298758-298758 on Build GRCh38
rs182943200 Chr.11: 298879-298879 on Build GRCh38
rs374540672 Chr.11: 298694-298694 on Build GRCh38
rs768779464 Chr.11: 298710-298710 on Build GRCh38
rs780000312 Chr.11: 298691-298691 on Build GRCh38
rs574425931 Chr.11: 298769-298769 on Build GRCh38
rs147113548 Chr.11: 298704-298704 on Build GRCh38
rs764997281 Chr.11: 298737-298737 on Build GRCh38
rs750449912 Chr.11: 298749-298749 on Build GRCh38
rs535849180 Chr.11: 298744-298744 on Build GRCh38
rs1004936171 Chr.11: 298702-298702 on Build GRCh38
rs898900636 Chr.11: 298707-298707 on Build GRCh38
rs190604142 Chr.11: 298777-298777 on Build GRCh38
rs868223964 Chr.11: 298784-298784 on Build GRCh38
rs772785846 Chr.11: 298714-298714 on Build GRCh38
rs745950756 Chr.11: 298679-298679 on Build GRCh38

View Full Product Details