Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94393408-94393681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR83
Entrez Gene ID
10888
Gene Name
G protein-coupled receptor 83
Gene Aliases
GIR, GPR72
UniGene
Hs.272385
Gene Chromosome Location
Chr.11: 94377311-94401479 on Build GRCh38
Transcripts
NM_016540.3 XM_011542559.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7149427 c.608C>A p.P203Q Chr.11: 94393524-94393524 on Build GRCh38
6519486 c.571G>T p.A191S Chr.11: 94393561-94393561 on Build GRCh38
212851 c.547A>G p.I183V Chr.11: 94393585-94393585 on Build GRCh38
6070770 c.543C>G p.I181M Chr.11: 94393589-94393589 on Build GRCh38
3454769 c.602C>T p.S201L Chr.11: 94393530-94393530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749950798 Chr.11: 94393429-94393429 on Build GRCh38
rs148456753 Chr.11: 94393566-94393566 on Build GRCh38
rs529436452 Chr.11: 94393587-94393587 on Build GRCh38
rs138745421 Chr.11: 94393538-94393538 on Build GRCh38
rs764146016 Chr.11: 94393501-94393501 on Build GRCh38
rs763041836 Chr.11: 94393494-94393494 on Build GRCh38
rs200169302 Chr.11: 94393451-94393451 on Build GRCh38
rs757785827 Chr.11: 94393634-94393634 on Build GRCh38
rs78029801 Chr.11: 94393454-94393454 on Build GRCh38
rs748734420 Chr.11: 94393459-94393459 on Build GRCh38
rs1047161078 Chr.11: 94393597-94393597 on Build GRCh38
rs746152073 Chr.11: 94393577-94393577 on Build GRCh38
rs770065005 Chr.11: 94393486-94393486 on Build GRCh38
rs764054129 Chr.11: 94393434-94393434 on Build GRCh38
rs1021102211 Chr.11: 94393556-94393556 on Build GRCh38
rs754533996 Chr.11: 94393611-94393611 on Build GRCh38
rs759789727 Chr.11: 94393479-94393479 on Build GRCh38
rs753457922 Chr.11: 94393609-94393609 on Build GRCh38
rs771296658 Chr.11: 94393471-94393471 on Build GRCh38
rs200007274 Chr.11: 94393455-94393455 on Build GRCh38
rs778205077 Chr.11: 94393461-94393461 on Build GRCh38
rs764792630 Chr.11: 94393614-94393614 on Build GRCh38
rs199882294 Chr.11: 94393593-94393593 on Build GRCh38
rs779539547 Chr.11: 94393457-94393457 on Build GRCh38
rs763066807 Chr.11: 94393497-94393497 on Build GRCh38
rs559424905 Chr.11: 94393654-94393654 on Build GRCh38
rs375569079 Chr.11: 94393589-94393589 on Build GRCh38
rs569609499 Chr.11: 94393584-94393584 on Build GRCh38
rs1031950129 Chr.11: 94393516-94393516 on Build GRCh38
rs1016467000 Chr.11: 94393494-94393494 on Build GRCh38
rs747365901 Chr.11: 94393470-94393470 on Build GRCh38
rs760416828 Chr.11: 94393605-94393605 on Build GRCh38
rs767317752 Chr.11: 94393523-94393523 on Build GRCh38
rs780459201 Chr.11: 94393641-94393641 on Build GRCh38
rs561456010 Chr.11: 94393583-94393583 on Build GRCh38
rs750114097 Chr.11: 94393531-94393531 on Build GRCh38
rs188775750 Chr.11: 94393655-94393655 on Build GRCh38
rs141669036 Chr.11: 94393585-94393585 on Build GRCh38
rs777784281 Chr.11: 94393601-94393601 on Build GRCh38
rs146239967 Chr.11: 94393562-94393562 on Build GRCh38
rs775714430 Chr.11: 94393496-94393496 on Build GRCh38
rs141900602 Chr.11: 94393561-94393561 on Build GRCh38
rs1005711419 Chr.11: 94393490-94393490 on Build GRCh38
rs996508667 Chr.11: 94393512-94393512 on Build GRCh38
rs771259350 Chr.11: 94393564-94393564 on Build GRCh38
rs767295508 Chr.11: 94393595-94393595 on Build GRCh38
rs192469914 Chr.11: 94393539-94393539 on Build GRCh38
rs202212504 Chr.11: 94393613-94393613 on Build GRCh38
rs776767658 Chr.11: 94393472-94393472 on Build GRCh38
rs773038473 Chr.11: 94393438-94393440 on Build GRCh38
rs751229373 Chr.11: 94393521-94393521 on Build GRCh38
rs749228835 Chr.11: 94393637-94393637 on Build GRCh38
rs757024701 Chr.11: 94393448-94393448 on Build GRCh38
rs751411906 Chr.11: 94393440-94393440 on Build GRCh38
rs987923818 Chr.11: 94393551-94393551 on Build GRCh38
rs751471958 Chr.11: 94393502-94393502 on Build GRCh38
rs772584375 Chr.11: 94393460-94393460 on Build GRCh38
rs201057163 Chr.11: 94393608-94393608 on Build GRCh38
rs376130003 Chr.11: 94393430-94393430 on Build GRCh38
rs201101741 Chr.11: 94393616-94393616 on Build GRCh38
rs755800073 Chr.11: 94393536-94393536 on Build GRCh38
rs200379878 Chr.11: 94393635-94393635 on Build GRCh38
rs372209016 Chr.11: 94393504-94393504 on Build GRCh38
rs200401802 Chr.11: 94393594-94393594 on Build GRCh38

View Full Product Details