Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157805725-157805974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLINT1
Entrez Gene ID
9685
Gene Name
clathrin interactor 1
Gene Aliases
CLINT, ENTH, EPN4, EPNR
UniGene
Hs.644000
Gene Chromosome Location
Chr.5: 157785743-157859175 on Build GRCh38
Transcripts
NM_001195555.1 NM_001195556.1 NM_014666.3 XM_011534701.2 XM_017010087.1 XM_017010088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4850949 c.878C>G p.S293C Chr.5: 157805876-157805876 on Build GRCh38
6170397 c.860C>G p.S287* Chr.5: 157805894-157805894 on Build GRCh38
4346238 c.875A>G p.H292R Chr.5: 157805933-157805933 on Build GRCh38
4672809 c.882A>G p.T294T Chr.5: 157805926-157805926 on Build GRCh38
6630333 c.886G>A p.D296N Chr.5: 157805922-157805922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891247034 Chr.5: 157805790-157805792 on Build GRCh38
rs762956245 Chr.5: 157805941-157805941 on Build GRCh38
rs768782531 Chr.5: 157805923-157805923 on Build GRCh38
rs748804538 Chr.5: 157805882-157805882 on Build GRCh38
rs751893699 Chr.5: 157805827-157805827 on Build GRCh38
rs372292185 Chr.5: 157805898-157805898 on Build GRCh38
rs766568926 Chr.5: 157805819-157805819 on Build GRCh38
rs1047781047 Chr.5: 157805814-157805814 on Build GRCh38
rs930743311 Chr.5: 157805802-157805802 on Build GRCh38
rs746914389 Chr.5: 157805905-157805905 on Build GRCh38
rs375245097 Chr.5: 157805888-157805888 on Build GRCh38
rs777198195 Chr.5: 157805842-157805842 on Build GRCh38
rs34237688 Chr.5: 157805925-157805926 on Build GRCh38
rs753228377 Chr.5: 157805844-157805844 on Build GRCh38
rs544549394 Chr.5: 157805927-157805927 on Build GRCh38
rs371784310 Chr.5: 157805843-157805843 on Build GRCh38
rs766255334 Chr.5: 157805791-157805791 on Build GRCh38
rs10515755 Chr.5: 157805756-157805756 on Build GRCh38
rs778430405 Chr.5: 157805884-157805884 on Build GRCh38
rs756873124 Chr.5: 157805883-157805883 on Build GRCh38
rs368145773 Chr.5: 157805891-157805891 on Build GRCh38
rs756664963 Chr.5: 157805845-157805845 on Build GRCh38
rs773329569 Chr.5: 157805933-157805933 on Build GRCh38
rs752803235 Chr.5: 157805793-157805793 on Build GRCh38
rs886465898 Chr.5: 157805815-157805815 on Build GRCh38
rs1007078813 Chr.5: 157805813-157805813 on Build GRCh38
rs777258160 Chr.5: 157805850-157805850 on Build GRCh38

View Full Product Details