Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 58160059-58160294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCL
Entrez Gene ID
55120
Gene Name
Fanconi anemia complementation group L
Gene Aliases
FAAP43, PHF9, POG
UniGene
Hs.631890
Gene Chromosome Location
Chr.2: 58159243-58241380 on Build GRCh38
Transcripts
NM_001114636.1 NM_018062.3 XM_005264395.3 XM_005264397.4 XM_011532939.2 XM_011532940.2 XM_011532941.1 XM_011532942.2 XM_011532944.2 XM_011532945.2 XM_017004414.1 XM_017004416.1 XM_005264540.4 XM_006712090.3 XM_006712091.3 XM_006712092.3 XM_006712093.3 XM_011533092.2 XM_017004858.1 XM_017004859.1 XM_017004860.1 XM_017004861.1 XM_017004862.1 XM_017004863.1 XM_017004864.1 XM_017004865.1 XM_017004866.1

Gene Symbol
VRK2
Entrez Gene ID
7444
Gene Name
vaccinia related kinase 2
Gene Aliases
-
UniGene
Hs.715298
Gene Chromosome Location
Chr.2: 57907651-58163991 on Build GRCh38
Transcripts
NM_001114636.1 NM_018062.3 XM_005264395.3 XM_005264397.4 XM_011532939.2 XM_011532940.2 XM_011532941.1 XM_011532942.2 XM_011532944.2 XM_011532945.2 XM_017004414.1 XM_017004416.1 XM_005264540.4 XM_006712090.3 XM_006712091.3 XM_006712092.3 XM_006712093.3 XM_011533092.2 XM_017004858.1 XM_017004859.1 XM_017004860.1 XM_017004861.1 XM_017004862.1 XM_017004863.1 XM_017004864.1 XM_017004865.1 XM_017004866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4991775 c.1026G>A p.L342L Chr.2: 58160174-58160174 on Build GRCh38
5019390 c.1077T>C p.C359C Chr.2: 58160123-58160123 on Build GRCh38
6422783 c.1087A>T p.S363C Chr.2: 58160113-58160113 on Build GRCh38
1614983 c.1049A>T p.Q350L Chr.2: 58160151-58160151 on Build GRCh38
5030061 c.1092+5G>C p.? Chr.2: 58160103-58160103 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765729177 Chr.2: 58160148-58160149 on Build GRCh38
rs140758986 Chr.2: 58160176-58160176 on Build GRCh38
rs933913059 Chr.2: 58160128-58160128 on Build GRCh38
rs763840016 Chr.2: 58160203-58160203 on Build GRCh38
rs758914385 Chr.2: 58160195-58160195 on Build GRCh38
rs768303607 Chr.2: 58160215-58160215 on Build GRCh38
rs147860433 Chr.2: 58160135-58160135 on Build GRCh38
rs377052216 Chr.2: 58160185-58160185 on Build GRCh38
rs766758036 Chr.2: 58160158-58160158 on Build GRCh38
rs941825695 Chr.2: 58160231-58160231 on Build GRCh38
rs909075867 Chr.2: 58160200-58160202 on Build GRCh38
rs928547539 Chr.2: 58160165-58160165 on Build GRCh38
rs898222946 Chr.2: 58160245-58160245 on Build GRCh38
rs370283793 Chr.2: 58160149-58160149 on Build GRCh38
rs371524685 Chr.2: 58160209-58160209 on Build GRCh38
rs778297632 Chr.2: 58160216-58160216 on Build GRCh38
rs375504895 Chr.2: 58160180-58160180 on Build GRCh38
rs750871999 Chr.2: 58160147-58160148 on Build GRCh38
rs777947589 Chr.2: 58160178-58160178 on Build GRCh38
rs760542163 Chr.2: 58160150-58160150 on Build GRCh38
rs773869051 Chr.2: 58160133-58160133 on Build GRCh38
rs994983497 Chr.2: 58160248-58160248 on Build GRCh38
rs747428014 Chr.2: 58160207-58160207 on Build GRCh38
rs577063114 Chr.2: 58160108-58160108 on Build GRCh38
rs971152606 Chr.2: 58160140-58160142 on Build GRCh38
rs11539575 Chr.2: 58160123-58160123 on Build GRCh38
rs181502442 Chr.2: 58160213-58160213 on Build GRCh38
rs778127243 Chr.2: 58160094-58160094 on Build GRCh38
rs753977997 Chr.2: 58160168-58160168 on Build GRCh38
rs374143605 Chr.2: 58160211-58160211 on Build GRCh38
rs544425912 Chr.2: 58160112-58160112 on Build GRCh38
rs748875030 Chr.2: 58160122-58160122 on Build GRCh38
rs774989597 Chr.2: 58160113-58160113 on Build GRCh38
rs778252009 Chr.2: 58160197-58160197 on Build GRCh38
rs373854636 Chr.2: 58160090-58160090 on Build GRCh38
rs766440818 Chr.2: 58160193-58160194 on Build GRCh38
rs145260146 Chr.2: 58160148-58160148 on Build GRCh38
rs929280865 Chr.2: 58160167-58160167 on Build GRCh38
rs763041336 Chr.2: 58160162-58160164 on Build GRCh38
rs760502912 Chr.2: 58160136-58160138 on Build GRCh38
rs747594960 Chr.2: 58160221-58160221 on Build GRCh38

View Full Product Details