Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154205443-154205704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf189
Entrez Gene ID
388701
Gene Name
chromosome 1 open reading frame 189
Gene Aliases
-
UniGene
Hs.104967
Gene Chromosome Location
Chr.1: 154199085-154206365 on Build GRCh38
Transcripts
NM_001010979.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
230459 c.58A>T p.K20* Chr.1: 154205614-154205614 on Build GRCh38
463043 c.57T>C p.L19L Chr.1: 154205615-154205615 on Build GRCh38
1626464 c.67G>T p.V23F Chr.1: 154205605-154205605 on Build GRCh38
6498089 c.70G>A p.D24N Chr.1: 154205602-154205602 on Build GRCh38
3750383 c.69C>T p.V23V Chr.1: 154205603-154205603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771302519 Chr.1: 154205565-154205565 on Build GRCh38
rs746365632 Chr.1: 154205654-154205654 on Build GRCh38
rs752438026 Chr.1: 154205627-154205627 on Build GRCh38
rs187876376 Chr.1: 154205667-154205667 on Build GRCh38
rs375288005 Chr.1: 154205584-154205584 on Build GRCh38
rs767856780 Chr.1: 154205620-154205620 on Build GRCh38
rs368277599 Chr.1: 154205606-154205606 on Build GRCh38
rs1031076519 Chr.1: 154205475-154205475 on Build GRCh38
rs749823963 Chr.1: 154205554-154205554 on Build GRCh38
rs202243041 Chr.1: 154205610-154205610 on Build GRCh38
rs988621533 Chr.1: 154205493-154205493 on Build GRCh38
rs779258827 Chr.1: 154205556-154205557 on Build GRCh38
rs370614128 Chr.1: 154205581-154205581 on Build GRCh38
rs772464237 Chr.1: 154205660-154205660 on Build GRCh38
rs554005927 Chr.1: 154205501-154205501 on Build GRCh38
rs777117624 Chr.1: 154205547-154205547 on Build GRCh38
rs74957241 Chr.1: 154205603-154205603 on Build GRCh38
rs940028116 Chr.1: 154205525-154205525 on Build GRCh38
rs1037041396 Chr.1: 154205530-154205530 on Build GRCh38
rs755305410 Chr.1: 154205637-154205638 on Build GRCh38
rs775753798 Chr.1: 154205589-154205589 on Build GRCh38
rs780372453 Chr.1: 154205663-154205663 on Build GRCh38
rs372535050 Chr.1: 154205622-154205622 on Build GRCh38
rs201820451 Chr.1: 154205639-154205639 on Build GRCh38
rs753219412 Chr.1: 154205646-154205646 on Build GRCh38
rs746455211 Chr.1: 154205608-154205610 on Build GRCh38
rs776780917 Chr.1: 154205675-154205675 on Build GRCh38
rs998208092 Chr.1: 154205473-154205473 on Build GRCh38
rs759464443 Chr.1: 154205623-154205623 on Build GRCh38
rs779252123 Chr.1: 154205576-154205576 on Build GRCh38
rs537411509 Chr.1: 154205672-154205672 on Build GRCh38
rs942900864 Chr.1: 154205637-154205637 on Build GRCh38
rs780396891 Chr.1: 154205610-154205613 on Build GRCh38
rs772469118 Chr.1: 154205610-154205610 on Build GRCh38
rs144170950 Chr.1: 154205607-154205607 on Build GRCh38
rs148215789 Chr.1: 154205605-154205605 on Build GRCh38
rs750767123 Chr.1: 154205625-154205625 on Build GRCh38
rs956787233 Chr.1: 154205485-154205485 on Build GRCh38
rs757795963 Chr.1: 154205650-154205650 on Build GRCh38

View Full Product Details