Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13889608-13889853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf57
Entrez Gene ID
79173
Gene Name
chromosome 19 open reading frame 57
Gene Aliases
-
UniGene
Hs.143288
Gene Chromosome Location
Chr.19: 13882348-13906125 on Build GRCh38
Transcripts
NM_024323.3 XM_006722885.3 XM_011528286.2 XM_011528287.2 XM_017027300.1 XM_017027301.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1685160 c.1247C>T p.T416I Chr.19: 13889609-13889609 on Build GRCh38
3692401 c.1077C>T p.P359P Chr.19: 13889779-13889779 on Build GRCh38
6614940 c.1003G>A p.G335R Chr.19: 13889853-13889853 on Build GRCh38
3529256 c.1242C>T p.G414G Chr.19: 13889614-13889614 on Build GRCh38
4074711 c.1057G>A p.A353T Chr.19: 13889799-13889799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751843990 Chr.19: 13889774-13889774 on Build GRCh38
rs145714071 Chr.19: 13889637-13889637 on Build GRCh38
rs757296683 Chr.19: 13889749-13889749 on Build GRCh38
rs371257595 Chr.19: 13889731-13889731 on Build GRCh38
rs755071844 Chr.19: 13889655-13889655 on Build GRCh38
rs763581645 Chr.19: 13889631-13889631 on Build GRCh38
rs748337980 Chr.19: 13889664-13889664 on Build GRCh38
rs377380467 Chr.19: 13889629-13889629 on Build GRCh38
rs755884538 Chr.19: 13889642-13889642 on Build GRCh38
rs778908609 Chr.19: 13889662-13889662 on Build GRCh38
rs747605182 Chr.19: 13889800-13889800 on Build GRCh38
rs771802249 Chr.19: 13889730-13889730 on Build GRCh38
rs766143368 Chr.19: 13889733-13889733 on Build GRCh38
rs770052483 Chr.19: 13889778-13889779 on Build GRCh38
rs753790903 Chr.19: 13889744-13889744 on Build GRCh38
rs772996823 Chr.19: 13889816-13889816 on Build GRCh38
rs202100982 Chr.19: 13889732-13889732 on Build GRCh38
rs146218274 Chr.19: 13889690-13889690 on Build GRCh38
rs777901983 Chr.19: 13889772-13889772 on Build GRCh38
rs761440547 Chr.19: 13889716-13889716 on Build GRCh38
rs774049459 Chr.19: 13889696-13889696 on Build GRCh38
rs145637934 Chr.19: 13889798-13889798 on Build GRCh38
rs780252706 Chr.19: 13889818-13889818 on Build GRCh38
rs1002452658 Chr.19: 13889658-13889658 on Build GRCh38
rs954613914 Chr.19: 13889793-13889793 on Build GRCh38
rs780728937 Chr.19: 13889785-13889785 on Build GRCh38
rs760291585 Chr.19: 13889826-13889826 on Build GRCh38
rs142928199 Chr.19: 13889654-13889654 on Build GRCh38
rs35608314 Chr.19: 13889631-13889632 on Build GRCh38
rs368853141 Chr.19: 13889779-13889779 on Build GRCh38
rs765190651 Chr.19: 13889757-13889757 on Build GRCh38
rs370798344 Chr.19: 13889632-13889632 on Build GRCh38
rs148917583 Chr.19: 13889778-13889778 on Build GRCh38
rs752721234 Chr.19: 13889763-13889763 on Build GRCh38
rs983428153 Chr.19: 13889803-13889803 on Build GRCh38
rs964616644 Chr.19: 13889760-13889760 on Build GRCh38
rs907904775 Chr.19: 13889815-13889815 on Build GRCh38
rs777989654 Chr.19: 13889683-13889683 on Build GRCh38
rs747313593 Chr.19: 13889689-13889689 on Build GRCh38
rs766283631 Chr.19: 13889646-13889646 on Build GRCh38
rs573709719 Chr.19: 13889692-13889692 on Build GRCh38
rs777015323 Chr.19: 13889628-13889628 on Build GRCh38
rs577821812 Chr.19: 13889693-13889693 on Build GRCh38
rs1026203416 Chr.19: 13889686-13889686 on Build GRCh38
rs758575040 Chr.19: 13889667-13889667 on Build GRCh38
rs776803571 Chr.19: 13889684-13889684 on Build GRCh38
rs758482140 Chr.19: 13889767-13889767 on Build GRCh38
rs759430899 Chr.19: 13889755-13889755 on Build GRCh38
rs372180863 Chr.19: 13889812-13889812 on Build GRCh38
rs781462058 Chr.19: 13889775-13889775 on Build GRCh38

View Full Product Details