Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123194722-123194947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLMP
Entrez Gene ID
79827
Gene Name
CXADR like membrane protein
Gene Aliases
ACAM, ASAM, CSBM, CSBS
UniGene
Hs.591949
Gene Chromosome Location
Chr.11: 123072006-123195305 on Build GRCh38
Transcripts
NM_024769.3 XM_011542998.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1704731 c.13C>T p.L5F Chr.11: 123194928-123194928 on Build GRCh38
1973627 c.5C>T p.S2F Chr.11: 123194936-123194936 on Build GRCh38
6131513 c.18C>A p.L6L Chr.11: 123194923-123194923 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs77817614 Chr.11: 123194752-123194752 on Build GRCh38
rs761506052 Chr.11: 123194923-123194930 on Build GRCh38
rs267602743 Chr.11: 123194913-123194913 on Build GRCh38
rs764915660 Chr.11: 123194924-123194926 on Build GRCh38
rs926154258 Chr.11: 123194883-123194883 on Build GRCh38
rs751310397 Chr.11: 123194877-123194877 on Build GRCh38
rs372397144 Chr.11: 123194890-123194890 on Build GRCh38
rs1018215560 Chr.11: 123194777-123194777 on Build GRCh38
rs1048262956 Chr.11: 123194762-123194762 on Build GRCh38
rs994398592 Chr.11: 123194760-123194760 on Build GRCh38
rs998197570 Chr.11: 123194816-123194816 on Build GRCh38
rs201793940 Chr.11: 123194879-123194879 on Build GRCh38
rs779162563 Chr.11: 123194924-123194924 on Build GRCh38
rs540076009 Chr.11: 123194773-123194773 on Build GRCh38
rs200559462 Chr.11: 123194866-123194866 on Build GRCh38
rs537803577 Chr.11: 123194793-123194793 on Build GRCh38
rs965346798 Chr.11: 123194815-123194815 on Build GRCh38
rs1055770918 Chr.11: 123194840-123194840 on Build GRCh38
rs780856345 Chr.11: 123194893-123194893 on Build GRCh38
rs556502081 Chr.11: 123194799-123194799 on Build GRCh38
rs762778910 Chr.11: 123194867-123194867 on Build GRCh38
rs752168769 Chr.11: 123194895-123194895 on Build GRCh38
rs753335389 Chr.11: 123194920-123194922 on Build GRCh38
rs989661428 Chr.11: 123194860-123194877 on Build GRCh38
rs763662657 Chr.11: 123194867-123194868 on Build GRCh38
rs115214371 Chr.11: 123194876-123194876 on Build GRCh38
rs888346250 Chr.11: 123194767-123194767 on Build GRCh38
rs757750315 Chr.11: 123194921-123194921 on Build GRCh38
rs370608003 Chr.11: 123194923-123194925 on Build GRCh38
rs1042568871 Chr.11: 123194763-123194763 on Build GRCh38
rs749804569 Chr.11: 123194912-123194912 on Build GRCh38
rs375458344 Chr.11: 123194904-123194904 on Build GRCh38
rs778461979 Chr.11: 123194906-123194906 on Build GRCh38

View Full Product Details