Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18008772-18009023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC2
Entrez Gene ID
27106
Gene Name
arrestin domain containing 2
Gene Aliases
CLONE24945, PP2703
UniGene
Hs.515249
Gene Chromosome Location
Chr.19: 18001132-18014102 on Build GRCh38
Transcripts
NM_001025604.2 NM_001286826.1 NM_015683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1391606 c.359G>A p.G120D Chr.19: 18009003-18009003 on Build GRCh38
6605232 c.358G>A p.G120S Chr.19: 18009002-18009002 on Build GRCh38
6999098 c.342-2A>G p.? Chr.19: 18008969-18008969 on Build GRCh38
6605229 c.379A>G p.K127E Chr.19: 18009023-18009023 on Build GRCh38
1391605 c.374G>A p.G125D Chr.19: 18009003-18009003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557279508 Chr.19: 18008884-18008884 on Build GRCh38
rs761957491 Chr.19: 18008967-18008967 on Build GRCh38
rs1030991008 Chr.19: 18008882-18008882 on Build GRCh38
rs373516769 Chr.19: 18008792-18008792 on Build GRCh38
rs751849884 Chr.19: 18008805-18008805 on Build GRCh38
rs752562062 Chr.19: 18008924-18008924 on Build GRCh38
rs754788726 Chr.19: 18008827-18008827 on Build GRCh38
rs535491434 Chr.19: 18008834-18008834 on Build GRCh38
rs768885081 Chr.19: 18008958-18008958 on Build GRCh38
rs778782166 Chr.19: 18008922-18008922 on Build GRCh38
rs890118153 Chr.19: 18008889-18008889 on Build GRCh38
rs770323668 Chr.19: 18008879-18008879 on Build GRCh38
rs144002717 Chr.19: 18008970-18008970 on Build GRCh38
rs988158329 Chr.19: 18008955-18008955 on Build GRCh38
rs577005786 Chr.19: 18008893-18008893 on Build GRCh38
rs148226511 Chr.19: 18008974-18008974 on Build GRCh38
rs943728105 Chr.19: 18008966-18008966 on Build GRCh38
rs1027158725 Chr.19: 18008939-18008942 on Build GRCh38
rs199938513 Chr.19: 18008800-18008800 on Build GRCh38
rs112781966 Chr.19: 18008859-18008859 on Build GRCh38
rs753622086 Chr.19: 18008820-18008820 on Build GRCh38
rs2305738 Chr.19: 18008869-18008869 on Build GRCh38
rs758372972 Chr.19: 18008985-18008985 on Build GRCh38
rs377380894 Chr.19: 18008809-18008809 on Build GRCh38
rs555763184 Chr.19: 18008856-18008856 on Build GRCh38
rs767744605 Chr.19: 18008819-18008819 on Build GRCh38
rs746492598 Chr.19: 18008932-18008932 on Build GRCh38
rs147287231 Chr.19: 18008971-18008971 on Build GRCh38
rs772996273 Chr.19: 18008795-18008824 on Build GRCh38
rs146526569 Chr.19: 18008894-18008894 on Build GRCh38
rs775877444 Chr.19: 18008948-18008948 on Build GRCh38
rs377430088 Chr.19: 18008959-18008959 on Build GRCh38
rs763508178 Chr.19: 18008956-18008956 on Build GRCh38
rs760450651 Chr.19: 18008801-18008811 on Build GRCh38
rs777227232 Chr.19: 18008930-18008930 on Build GRCh38
rs973657078 Chr.19: 18008898-18008898 on Build GRCh38
rs574806528 Chr.19: 18008858-18008858 on Build GRCh38
rs370560777 Chr.19: 18008925-18008925 on Build GRCh38
rs566910257 Chr.19: 18008814-18008814 on Build GRCh38
rs189196235 Chr.19: 18008992-18008992 on Build GRCh38
rs12609555 Chr.19: 18008942-18008942 on Build GRCh38
rs373925694 Chr.19: 18008804-18008804 on Build GRCh38
rs868649991 Chr.19: 18008892-18008892 on Build GRCh38
rs866578224 Chr.19: 18008895-18008895 on Build GRCh38
rs751130030 Chr.19: 18009001-18009001 on Build GRCh38
rs1027805629 Chr.19: 18008917-18008917 on Build GRCh38
rs868570374 Chr.19: 18008969-18008969 on Build GRCh38
rs12610703 Chr.19: 18008946-18008946 on Build GRCh38
rs184883492 Chr.19: 18008944-18008944 on Build GRCh38
rs752664056 Chr.19: 18008977-18008977 on Build GRCh38

View Full Product Details